Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30722825-30723098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUBB
Entrez Gene ID
203068
Gene Name
tubulin beta class I
Gene Aliases
CDCBM6, CSCSC1, M40, OK/SW-cl.56, TUBB1, TUBB5
UniGene
Hs.636480
Gene Chromosome Location
Chr.6: 30720201-30725426 on Build GRCh38
Transcripts
NM_001293212.1 NM_001293213.1 NM_001293214.1 NM_001293215.1 NM_001293216.1 NM_178014.3 NR_120608.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5501722 c.200A>G p.D67G Chr.6: 30722951-30722951 on Build GRCh38
3624363 c.238C>T p.P80S Chr.6: 30722989-30722989 on Build GRCh38
7287415 c.217A>G p.M73V Chr.6: 30722968-30722968 on Build GRCh38
6814025 c.221A>G p.D74G Chr.6: 30722972-30722972 on Build GRCh38
6019139 c.224C>G p.S75C Chr.6: 30722975-30722975 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774570416 Chr.6: 30722943-30722943 on Build GRCh38
rs777419483 Chr.6: 30722875-30722875 on Build GRCh38
rs752724910 Chr.6: 30722871-30722871 on Build GRCh38
rs770041203 Chr.6: 30722910-30722910 on Build GRCh38
rs757735804 Chr.6: 30723066-30723066 on Build GRCh38
rs25526 Chr.6: 30723039-30723039 on Build GRCh38
rs752154308 Chr.6: 30723048-30723048 on Build GRCh38
rs765121362 Chr.6: 30723042-30723042 on Build GRCh38
rs17189364 Chr.6: 30722858-30722858 on Build GRCh38
rs780301475 Chr.6: 30722912-30722912 on Build GRCh38
rs752205645 Chr.6: 30722872-30722872 on Build GRCh38
rs768761672 Chr.6: 30722931-30722931 on Build GRCh38
rs567974983 Chr.6: 30723051-30723051 on Build GRCh38
rs760458756 Chr.6: 30722946-30722946 on Build GRCh38
rs375422131 Chr.6: 30723056-30723056 on Build GRCh38
rs536531543 Chr.6: 30723057-30723057 on Build GRCh38
rs1020627067 Chr.6: 30722854-30722854 on Build GRCh38
rs770684802 Chr.6: 30723015-30723015 on Build GRCh38
rs781781352 Chr.6: 30722893-30722893 on Build GRCh38
rs781671861 Chr.6: 30723067-30723067 on Build GRCh38
rs757682952 Chr.6: 30722886-30722886 on Build GRCh38
rs776601686 Chr.6: 30723036-30723036 on Build GRCh38
rs1000538751 Chr.6: 30722860-30722860 on Build GRCh38
rs372823186 Chr.6: 30723043-30723043 on Build GRCh38
rs746344292 Chr.6: 30722900-30722900 on Build GRCh38
rs11546738 Chr.6: 30722947-30722947 on Build GRCh38

View Full Product Details