Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 32833136-32833365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC7A9
Entrez Gene ID
11136
Gene Name
solute carrier family 7 member 9
Gene Aliases
BAT1, CSNU3
UniGene
Hs.743345
Gene Chromosome Location
Chr.19: 32830511-32870958 on Build GRCh38
Transcripts
NM_001126335.1 NM_001243036.1 NM_014270.4 XM_006722992.1 XM_011526402.2 XM_017026230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1266052 c.1285A>G p.I429V Chr.19: 32833263-32833263 on Build GRCh38
994612 c.1373A>C p.K458T Chr.19: 32833175-32833175 on Build GRCh38
4914969 c.1233A>T p.V411V Chr.19: 32833315-32833315 on Build GRCh38
6387355 c.1383G>T p.W461C Chr.19: 32833165-32833165 on Build GRCh38
3532189 c.1379G>A p.G460E Chr.19: 32833169-32833169 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780783383 Chr.19: 32833207-32833207 on Build GRCh38
rs778565214 Chr.19: 32833340-32833340 on Build GRCh38
rs143213759 Chr.19: 32833294-32833294 on Build GRCh38
rs267605414 Chr.19: 32833308-32833308 on Build GRCh38
rs143999252 Chr.19: 32833305-32833305 on Build GRCh38
rs564489064 Chr.19: 32833170-32833170 on Build GRCh38
rs780106685 Chr.19: 32833219-32833219 on Build GRCh38
rs770047693 Chr.19: 32833293-32833293 on Build GRCh38
rs926998425 Chr.19: 32833328-32833328 on Build GRCh38
rs755978655 Chr.19: 32833334-32833334 on Build GRCh38
rs913172054 Chr.19: 32833281-32833281 on Build GRCh38
rs747049300 Chr.19: 32833251-32833251 on Build GRCh38
rs779653925 Chr.19: 32833281-32833282 on Build GRCh38
rs767216278 Chr.19: 32833330-32833330 on Build GRCh38
rs753324280 Chr.19: 32833244-32833244 on Build GRCh38
rs771287087 Chr.19: 32833258-32833258 on Build GRCh38
rs965239146 Chr.19: 32833231-32833231 on Build GRCh38
rs199552160 Chr.19: 32833210-32833210 on Build GRCh38
rs753692696 Chr.19: 32833285-32833286 on Build GRCh38
rs763478661 Chr.19: 32833178-32833178 on Build GRCh38
rs750233159 Chr.19: 32833209-32833209 on Build GRCh38
rs536084711 Chr.19: 32833306-32833306 on Build GRCh38
rs34347941 Chr.19: 32833183-32833183 on Build GRCh38
rs749151193 Chr.19: 32833223-32833223 on Build GRCh38
rs200721127 Chr.19: 32833167-32833167 on Build GRCh38
rs374629909 Chr.19: 32833331-32833331 on Build GRCh38
rs1054280997 Chr.19: 32833259-32833259 on Build GRCh38
rs988712826 Chr.19: 32833242-32833242 on Build GRCh38
rs760033221 Chr.19: 32833272-32833272 on Build GRCh38
rs776932048 Chr.19: 32833261-32833261 on Build GRCh38
rs370170089 Chr.19: 32833318-32833318 on Build GRCh38
rs1007096305 Chr.19: 32833195-32833195 on Build GRCh38
rs138086959 Chr.19: 32833179-32833179 on Build GRCh38
rs139998987 Chr.19: 32833317-32833317 on Build GRCh38
rs754996602 Chr.19: 32833314-32833315 on Build GRCh38
rs772245267 Chr.19: 32833237-32833237 on Build GRCh38

View Full Product Details