Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 114356018-114356243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX5
Entrez Gene ID
6910
Gene Name
T-box 5
Gene Aliases
HOS
UniGene
Hs.381715
Gene Chromosome Location
Chr.12: 114353911-114408708 on Build GRCh38
Transcripts
NM_000192.3 NM_080717.2 NM_181486.2 XM_017019912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5901824 c.1055C>T p.S352F Chr.12: 114356034-114356034 on Build GRCh38
6798000 c.1064G>A p.R355H Chr.12: 114356025-114356025 on Build GRCh38
6071340 c.1006C>T p.H336Y Chr.12: 114356083-114356083 on Build GRCh38
4732560 c.1063C>T p.R355C Chr.12: 114356026-114356026 on Build GRCh38
6135722 c.1039C>G p.P347A Chr.12: 114356050-114356050 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907617752 Chr.12: 114356068-114356068 on Build GRCh38
rs760830193 Chr.12: 114356074-114356074 on Build GRCh38
rs368472780 Chr.12: 114356114-114356114 on Build GRCh38
rs944423586 Chr.12: 114356079-114356079 on Build GRCh38
rs148977955 Chr.12: 114356069-114356069 on Build GRCh38
rs201105444 Chr.12: 114356118-114356118 on Build GRCh38
rs765327234 Chr.12: 114356098-114356098 on Build GRCh38
rs201816980 Chr.12: 114356113-114356113 on Build GRCh38
rs778011764 Chr.12: 114356050-114356050 on Build GRCh38
rs779437406 Chr.12: 114356149-114356149 on Build GRCh38
rs750350579 Chr.12: 114356105-114356105 on Build GRCh38
rs374446377 Chr.12: 114356094-114356094 on Build GRCh38
rs764322332 Chr.12: 114356076-114356076 on Build GRCh38
rs375002260 Chr.12: 114356121-114356121 on Build GRCh38
rs776297033 Chr.12: 114356085-114356085 on Build GRCh38
rs749598173 Chr.12: 114356055-114356055 on Build GRCh38
rs745997257 Chr.12: 114356061-114356061 on Build GRCh38
rs546488473 Chr.12: 114356204-114356204 on Build GRCh38
rs771408470 Chr.12: 114356057-114356057 on Build GRCh38
rs772109169 Chr.12: 114356067-114356067 on Build GRCh38
rs371873510 Chr.12: 114356119-114356119 on Build GRCh38
rs774825317 Chr.12: 114356060-114356060 on Build GRCh38
rs766007057 Chr.12: 114356116-114356116 on Build GRCh38
rs142807139 Chr.12: 114356104-114356104 on Build GRCh38
rs931630578 Chr.12: 114356127-114356127 on Build GRCh38
rs181923192 Chr.12: 114356136-114356136 on Build GRCh38
rs757596478 Chr.12: 114356145-114356145 on Build GRCh38

View Full Product Details