Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 45673602-45673831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SQRDL
Entrez Gene ID
58472
Gene Name
sulfide quinone reductase-like (yeast)
Gene Aliases
CGI-44, PRO1975, SQOR
UniGene
Hs.511251
Gene Chromosome Location
Chr.15: 45631148-45691294 on Build GRCh38
Transcripts
NM_001271213.1 NM_021199.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
470724 c.507T>G p.N169K Chr.15: 45673654-45673654 on Build GRCh38
3501738 c.555C>T p.F185F Chr.15: 45673702-45673702 on Build GRCh38
3501737 c.537G>A p.W179* Chr.15: 45673684-45673684 on Build GRCh38
5539708 c.641C>T p.A214V Chr.15: 45673788-45673788 on Build GRCh38
1640261 c.572T>C p.I191T Chr.15: 45673719-45673719 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546292950 Chr.15: 45673712-45673712 on Build GRCh38
rs765759874 Chr.15: 45673795-45673795 on Build GRCh38
rs143048388 Chr.15: 45673697-45673697 on Build GRCh38
rs149235309 Chr.15: 45673682-45673682 on Build GRCh38
rs754706953 Chr.15: 45673704-45673704 on Build GRCh38
rs764345186 Chr.15: 45673658-45673658 on Build GRCh38
rs749279381 Chr.15: 45673745-45673745 on Build GRCh38
rs765751532 Chr.15: 45673711-45673711 on Build GRCh38
rs772223842 Chr.15: 45673650-45673650 on Build GRCh38
rs769417350 Chr.15: 45673752-45673752 on Build GRCh38
rs762187581 Chr.15: 45673673-45673673 on Build GRCh38
rs142463548 Chr.15: 45673630-45673630 on Build GRCh38
rs955846683 Chr.15: 45673649-45673649 on Build GRCh38
rs776794288 Chr.15: 45673773-45673773 on Build GRCh38
rs768953493 Chr.15: 45673770-45673770 on Build GRCh38
rs760762286 Chr.15: 45673758-45673758 on Build GRCh38
rs928564640 Chr.15: 45673804-45673804 on Build GRCh38
rs80153459 Chr.15: 45673634-45673634 on Build GRCh38
rs761023993 Chr.15: 45673653-45673653 on Build GRCh38
rs754253932 Chr.15: 45673664-45673664 on Build GRCh38
rs769742146 Chr.15: 45673631-45673631 on Build GRCh38
rs1030478996 Chr.15: 45673641-45673641 on Build GRCh38
rs777701080 Chr.15: 45673743-45673743 on Build GRCh38
rs34450206 Chr.15: 45673649-45673650 on Build GRCh38
rs374794265 Chr.15: 45673797-45673797 on Build GRCh38
rs762256045 Chr.15: 45673784-45673784 on Build GRCh38
rs377509781 Chr.15: 45673747-45673747 on Build GRCh38
rs373774975 Chr.15: 45673680-45673680 on Build GRCh38
rs769651120 Chr.15: 45673734-45673734 on Build GRCh38
rs777578030 Chr.15: 45673758-45673758 on Build GRCh38
rs748002418 Chr.15: 45673718-45673718 on Build GRCh38
rs749040008 Chr.15: 45673773-45673774 on Build GRCh38
rs981343470 Chr.15: 45673791-45673791 on Build GRCh38
rs775674151 Chr.15: 45673651-45673651 on Build GRCh38
rs145954867 Chr.15: 45673633-45673633 on Build GRCh38
rs779780017 Chr.15: 45673696-45673696 on Build GRCh38
rs763597705 Chr.15: 45673798-45673798 on Build GRCh38
rs757970989 Chr.15: 45673693-45673693 on Build GRCh38

View Full Product Details