Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 53105518-53105789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFT1
Entrez Gene ID
91869
Gene Name
RFT1 homolog
Gene Aliases
CDG1N
UniGene
Hs.631910
Gene Chromosome Location
Chr.3: 53071151-53130469 on Build GRCh38
Transcripts
NM_052859.3 XM_005265537.4 XM_006713384.3 XM_011534214.2 XM_011534215.2 XM_011534216.2 XM_017007460.1 XM_017007461.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6034177 c.906_907insA p.F303fs*4 Chr.3: 53105723-53105724 on Build GRCh38
1424603 c.932G>A p.G311E Chr.3: 53105698-53105698 on Build GRCh38
2152019 c.882G>T p.Q294H Chr.3: 53105748-53105748 on Build GRCh38
6765302 c.855C>A p.G285G Chr.3: 53105775-53105775 on Build GRCh38
4983482 c.843G>C p.V281V Chr.3: 53105787-53105787 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376702575 Chr.3: 53105652-53105652 on Build GRCh38
rs938866151 Chr.3: 53105694-53105694 on Build GRCh38
rs948461251 Chr.3: 53105565-53105565 on Build GRCh38
rs777941484 Chr.3: 53105674-53105674 on Build GRCh38
rs75323042 Chr.3: 53105628-53105628 on Build GRCh38
rs755680371 Chr.3: 53105681-53105681 on Build GRCh38
rs913477149 Chr.3: 53105728-53105728 on Build GRCh38
rs773918760 Chr.3: 53105725-53105725 on Build GRCh38
rs796053521 Chr.3: 53105738-53105738 on Build GRCh38
rs766631068 Chr.3: 53105627-53105627 on Build GRCh38
rs140603849 Chr.3: 53105706-53105706 on Build GRCh38
rs552860074 Chr.3: 53105635-53105635 on Build GRCh38
rs759902313 Chr.3: 53105752-53105752 on Build GRCh38
rs886058724 Chr.3: 53105673-53105673 on Build GRCh38
rs947641452 Chr.3: 53105729-53105729 on Build GRCh38
rs779780383 Chr.3: 53105685-53105685 on Build GRCh38
rs768124396 Chr.3: 53105712-53105712 on Build GRCh38
rs752836352 Chr.3: 53105759-53105759 on Build GRCh38
rs760552260 Chr.3: 53105744-53105744 on Build GRCh38
rs765390877 Chr.3: 53105756-53105756 on Build GRCh38
rs372126941 Chr.3: 53105651-53105651 on Build GRCh38
rs770676975 Chr.3: 53105746-53105746 on Build GRCh38
rs900517283 Chr.3: 53105765-53105765 on Build GRCh38
rs748068081 Chr.3: 53105726-53105726 on Build GRCh38
rs201430920 Chr.3: 53105739-53105739 on Build GRCh38
rs747402128 Chr.3: 53105676-53105676 on Build GRCh38
rs890618317 Chr.3: 53105580-53105580 on Build GRCh38
rs753247463 Chr.3: 53105664-53105664 on Build GRCh38
rs760754137 Chr.3: 53105626-53105626 on Build GRCh38
rs776958467 Chr.3: 53105751-53105751 on Build GRCh38
rs570429921 Chr.3: 53105589-53105589 on Build GRCh38
rs919618712 Chr.3: 53105560-53105560 on Build GRCh38
rs772063119 Chr.3: 53105734-53105734 on Build GRCh38
rs759032044 Chr.3: 53105670-53105670 on Build GRCh38
rs772820136 Chr.3: 53105743-53105743 on Build GRCh38

View Full Product Details