Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 69895031-69895276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNRNP27
Entrez Gene ID
11017
Gene Name
small nuclear ribonucleoprotein U4/U6.U5 subunit 27
Gene Aliases
27K, RY1
UniGene
Hs.54649
Gene Chromosome Location
Chr.2: 69893943-69905236 on Build GRCh38
Transcripts
NM_006857.2 NR_037862.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5566285 c.148C>T p.R50C Chr.2: 69895207-69895207 on Build GRCh38
7089894 c.61C>A p.R21R Chr.2: 69895120-69895120 on Build GRCh38
7260366 c.101G>T p.R34L Chr.2: 69895160-69895160 on Build GRCh38
7275119 c.100C>T p.R34W Chr.2: 69895159-69895159 on Build GRCh38
6266221 c.50G>T p.R17L Chr.2: 69895109-69895109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758529525 Chr.2: 69895221-69895221 on Build GRCh38
rs11540988 Chr.2: 69895173-69895173 on Build GRCh38
rs1055209804 Chr.2: 69895103-69895103 on Build GRCh38
rs764031045 Chr.2: 69895113-69895113 on Build GRCh38
rs762819059 Chr.2: 69895105-69895105 on Build GRCh38
rs778883994 Chr.2: 69895058-69895058 on Build GRCh38
rs1046825252 Chr.2: 69895144-69895144 on Build GRCh38
rs1017125915 Chr.2: 69895256-69895256 on Build GRCh38
rs769444357 Chr.2: 69895093-69895093 on Build GRCh38
rs200831065 Chr.2: 69895122-69895122 on Build GRCh38
rs773563795 Chr.2: 69895084-69895084 on Build GRCh38
rs748758016 Chr.2: 69895230-69895234 on Build GRCh38
rs374836941 Chr.2: 69895116-69895116 on Build GRCh38
rs1037063859 Chr.2: 69895140-69895140 on Build GRCh38
rs546438538 Chr.2: 69895164-69895164 on Build GRCh38
rs200650012 Chr.2: 69895212-69895212 on Build GRCh38
rs143382169 Chr.2: 69895183-69895183 on Build GRCh38
rs551796621 Chr.2: 69895238-69895238 on Build GRCh38
rs141178820 Chr.2: 69895137-69895137 on Build GRCh38
rs773029374 Chr.2: 69895191-69895191 on Build GRCh38
rs978665536 Chr.2: 69895054-69895054 on Build GRCh38
rs774807862 Chr.2: 69895089-69895089 on Build GRCh38
rs968575180 Chr.2: 69895053-69895053 on Build GRCh38
rs748306160 Chr.2: 69895219-69895219 on Build GRCh38
rs927110163 Chr.2: 69895083-69895083 on Build GRCh38
rs772428730 Chr.2: 69895068-69895068 on Build GRCh38
rs775314643 Chr.2: 69895104-69895104 on Build GRCh38
rs750059083 Chr.2: 69895170-69895170 on Build GRCh38
rs182377309 Chr.2: 69895063-69895063 on Build GRCh38
rs755455254 Chr.2: 69895187-69895187 on Build GRCh38
rs778179995 Chr.2: 69895240-69895240 on Build GRCh38

View Full Product Details