Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30555970-30556233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF764
Entrez Gene ID
92595
Gene Name
zinc finger protein 764
Gene Aliases
-
UniGene
Hs.132227
Gene Chromosome Location
Chr.16: 30553764-30558321 on Build GRCh38
Transcripts
NM_001172679.1 NM_033410.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6832901 c.311-1G>A p.? Chr.16: 30556111-30556111 on Build GRCh38
1377412 c.434C>T p.P145L Chr.16: 30555987-30555987 on Build GRCh38
1377413 c.417C>T p.A139A Chr.16: 30556004-30556004 on Build GRCh38
5880457 c.388G>A p.G130R Chr.16: 30556033-30556033 on Build GRCh38
137641 c.422C>T p.S141L Chr.16: 30555999-30555999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749000414 Chr.16: 30556125-30556125 on Build GRCh38
rs767946886 Chr.16: 30556076-30556086 on Build GRCh38
rs1013973699 Chr.16: 30556175-30556175 on Build GRCh38
rs753218060 Chr.16: 30556090-30556097 on Build GRCh38
rs1039774615 Chr.16: 30556090-30556090 on Build GRCh38
rs199626772 Chr.16: 30556063-30556063 on Build GRCh38
rs550548880 Chr.16: 30556117-30556117 on Build GRCh38
rs904222867 Chr.16: 30556126-30556126 on Build GRCh38
rs777210908 Chr.16: 30556120-30556120 on Build GRCh38
rs773422797 Chr.16: 30556155-30556155 on Build GRCh38
rs772442020 Chr.16: 30556040-30556040 on Build GRCh38
rs754992430 Chr.16: 30556096-30556096 on Build GRCh38
rs752645117 Chr.16: 30556111-30556111 on Build GRCh38
rs183574944 Chr.16: 30556206-30556206 on Build GRCh38
rs760032356 Chr.16: 30556076-30556080 on Build GRCh38
rs890872682 Chr.16: 30556057-30556057 on Build GRCh38
rs929808234 Chr.16: 30556010-30556010 on Build GRCh38
rs748875795 Chr.16: 30556037-30556037 on Build GRCh38
rs538592097 Chr.16: 30556048-30556048 on Build GRCh38
rs149618421 Chr.16: 30556107-30556107 on Build GRCh38
rs370707272 Chr.16: 30556158-30556158 on Build GRCh38
rs137968351 Chr.16: 30556075-30556075 on Build GRCh38
rs775694517 Chr.16: 30556016-30556016 on Build GRCh38
rs754869498 Chr.16: 30556009-30556009 on Build GRCh38
rs374302641 Chr.16: 30555989-30555989 on Build GRCh38
rs750624227 Chr.16: 30556002-30556002 on Build GRCh38
rs1010366489 Chr.16: 30556164-30556164 on Build GRCh38
rs769009164 Chr.16: 30556049-30556049 on Build GRCh38
rs772886904 Chr.16: 30556034-30556034 on Build GRCh38
rs765367937 Chr.16: 30556067-30556067 on Build GRCh38
rs370974156 Chr.16: 30556141-30556141 on Build GRCh38
rs777035610 Chr.16: 30556146-30556146 on Build GRCh38
rs972824295 Chr.16: 30556032-30556032 on Build GRCh38
rs769249249 Chr.16: 30556019-30556019 on Build GRCh38
rs769016514 Chr.16: 30556142-30556142 on Build GRCh38
rs138651281 Chr.16: 30555991-30555991 on Build GRCh38
rs1024914802 Chr.16: 30556183-30556183 on Build GRCh38
rs753854899 Chr.16: 30555990-30555990 on Build GRCh38
rs547041727 Chr.16: 30556072-30556072 on Build GRCh38
rs776806681 Chr.16: 30556174-30556174 on Build GRCh38
rs377534613 Chr.16: 30556154-30556154 on Build GRCh38
rs751282369 Chr.16: 30556077-30556077 on Build GRCh38
rs747027870 Chr.16: 30556007-30556007 on Build GRCh38
rs776817455 Chr.16: 30556061-30556061 on Build GRCh38
rs368272428 Chr.16: 30555999-30555999 on Build GRCh38
rs780795395 Chr.16: 30556015-30556015 on Build GRCh38
rs769789684 Chr.16: 30556187-30556187 on Build GRCh38
rs879671670 Chr.16: 30556201-30556201 on Build GRCh38
rs763207824 Chr.16: 30556068-30556068 on Build GRCh38
rs115131694 Chr.16: 30556005-30556005 on Build GRCh38
rs903541729 Chr.16: 30556093-30556093 on Build GRCh38
rs548533265 Chr.16: 30556184-30556184 on Build GRCh38
rs770563942 Chr.16: 30556127-30556127 on Build GRCh38
rs191748145 Chr.16: 30556151-30556151 on Build GRCh38

View Full Product Details