Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 88831822-88832089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD22
Entrez Gene ID
118932
Gene Name
ankyrin repeat domain 22
Gene Aliases
-
UniGene
Hs.217484
Gene Chromosome Location
Chr.10: 88819899-88851975 on Build GRCh38
Transcripts
NM_144590.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4016475 c.139G>A p.A47T Chr.10: 88831909-88831909 on Build GRCh38
1702747 c.211C>T p.Q71* Chr.10: 88831837-88831837 on Build GRCh38
1492346 c.148C>T p.R50* Chr.10: 88831900-88831900 on Build GRCh38
3807928 c.74T>G p.V25G Chr.10: 88831974-88831974 on Build GRCh38
921205 c.28T>C p.C10R Chr.10: 88832020-88832020 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1004682746 Chr.10: 88831952-88831952 on Build GRCh38
rs367882066 Chr.10: 88831949-88831949 on Build GRCh38
rs761875744 Chr.10: 88831886-88831886 on Build GRCh38
rs761751044 Chr.10: 88831974-88831974 on Build GRCh38
rs368264244 Chr.10: 88832055-88832055 on Build GRCh38
rs767237871 Chr.10: 88832007-88832007 on Build GRCh38
rs201805869 Chr.10: 88832034-88832034 on Build GRCh38
rs772258711 Chr.10: 88831874-88831874 on Build GRCh38
rs768741256 Chr.10: 88832051-88832051 on Build GRCh38
rs565840632 Chr.10: 88832032-88832032 on Build GRCh38
rs1012105323 Chr.10: 88832024-88832024 on Build GRCh38
rs571700883 Chr.10: 88831923-88831923 on Build GRCh38
rs754616477 Chr.10: 88832001-88832001 on Build GRCh38
rs765816273 Chr.10: 88831984-88831984 on Build GRCh38
rs199828548 Chr.10: 88831902-88831902 on Build GRCh38
rs758113752 Chr.10: 88831977-88831977 on Build GRCh38
rs765255974 Chr.10: 88831976-88831976 on Build GRCh38
rs1023406640 Chr.10: 88831876-88831876 on Build GRCh38
rs200441199 Chr.10: 88831994-88831994 on Build GRCh38
rs747156601 Chr.10: 88832047-88832047 on Build GRCh38
rs886529179 Chr.10: 88831950-88831950 on Build GRCh38
rs201028486 Chr.10: 88831854-88831854 on Build GRCh38
rs372411272 Chr.10: 88832033-88832033 on Build GRCh38
rs745934666 Chr.10: 88831946-88831946 on Build GRCh38
rs947617486 Chr.10: 88831912-88831912 on Build GRCh38
rs765308788 Chr.10: 88831889-88831889 on Build GRCh38
rs377061999 Chr.10: 88831900-88831900 on Build GRCh38
rs922083908 Chr.10: 88831932-88831932 on Build GRCh38
rs146755980 Chr.10: 88831953-88831953 on Build GRCh38
rs559515763 Chr.10: 88831980-88831980 on Build GRCh38
rs749753273 Chr.10: 88831922-88831922 on Build GRCh38
rs779445573 Chr.10: 88831941-88831941 on Build GRCh38
rs370135851 Chr.10: 88831901-88831901 on Build GRCh38
rs115245413 Chr.10: 88832037-88832037 on Build GRCh38
rs754179745 Chr.10: 88832028-88832028 on Build GRCh38
rs775642379 Chr.10: 88831878-88831878 on Build GRCh38
rs759652210 Chr.10: 88831853-88831853 on Build GRCh38
rs760451952 Chr.10: 88831882-88831882 on Build GRCh38
rs374841412 Chr.10: 88831921-88831921 on Build GRCh38
rs776262103 Chr.10: 88831970-88831970 on Build GRCh38
rs117613865 Chr.10: 88831981-88831981 on Build GRCh38
rs772050443 Chr.10: 88831948-88831948 on Build GRCh38
rs768458768 Chr.10: 88831966-88831966 on Build GRCh38
rs749935435 Chr.10: 88831893-88831893 on Build GRCh38
rs542726939 Chr.10: 88831883-88831883 on Build GRCh38
rs1003379206 Chr.10: 88832041-88832041 on Build GRCh38
rs200891481 Chr.10: 88832020-88832020 on Build GRCh38
rs143516492 Chr.10: 88831899-88831899 on Build GRCh38
rs756440980 Chr.10: 88831917-88831917 on Build GRCh38
rs748104654 Chr.10: 88831993-88831994 on Build GRCh38

View Full Product Details