Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6478919-6479188 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFIP2
Entrez Gene ID
23647
Gene Name
ADP ribosylation factor interacting protein 2
Gene Aliases
POR1
UniGene
Hs.732203
Gene Chromosome Location
Chr.11: 6474683-6481479 on Build GRCh38
Transcripts
NM_001242854.1 NM_001242855.1 NM_001242856.1 NM_012402.3 XM_005252840.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1675798 c.311A>G p.Y104C Chr.11: 6479144-6479144 on Build GRCh38
7141921 c.341G>T p.R114L Chr.11: 6478934-6478934 on Build GRCh38
6036903 c.315G>A p.K105K Chr.11: 6479140-6479140 on Build GRCh38
6069887 c.313A>T p.K105* Chr.11: 6479142-6479142 on Build GRCh38
3451730 c.341G>A p.R114Q Chr.11: 6478934-6478934 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778856434 Chr.11: 6479100-6479100 on Build GRCh38
rs2253104 Chr.11: 6479079-6479079 on Build GRCh38
rs770112599 Chr.11: 6478957-6478957 on Build GRCh38
rs59266898 Chr.11: 6479034-6479034 on Build GRCh38
rs753493186 Chr.11: 6478993-6478993 on Build GRCh38
rs1021154893 Chr.11: 6479076-6479076 on Build GRCh38
rs755060844 Chr.11: 6479136-6479136 on Build GRCh38
rs1007788227 Chr.11: 6479018-6479018 on Build GRCh38
rs376630295 Chr.11: 6478973-6478973 on Build GRCh38
rs1039007252 Chr.11: 6479023-6479023 on Build GRCh38
rs967181836 Chr.11: 6479077-6479077 on Build GRCh38
rs760146169 Chr.11: 6478977-6478977 on Build GRCh38
rs767822111 Chr.11: 6478992-6478993 on Build GRCh38
rs768951451 Chr.11: 6478941-6478941 on Build GRCh38
rs371542802 Chr.11: 6478966-6478966 on Build GRCh38
rs540595840 Chr.11: 6479017-6479017 on Build GRCh38
rs745403178 Chr.11: 6479015-6479015 on Build GRCh38
rs200729824 Chr.11: 6479087-6479087 on Build GRCh38
rs763641373 Chr.11: 6478991-6478991 on Build GRCh38
rs886412445 Chr.11: 6479066-6479066 on Build GRCh38
rs760802374 Chr.11: 6479001-6479019 on Build GRCh38
rs367877916 Chr.11: 6479118-6479118 on Build GRCh38
rs890149161 Chr.11: 6479013-6479013 on Build GRCh38
rs762070550 Chr.11: 6478946-6478946 on Build GRCh38
rs75147856 Chr.11: 6479080-6479080 on Build GRCh38
rs1011074703 Chr.11: 6479044-6479044 on Build GRCh38
rs1047702717 Chr.11: 6479036-6479036 on Build GRCh38
rs746933394 Chr.11: 6479131-6479131 on Build GRCh38
rs868062670 Chr.11: 6479086-6479086 on Build GRCh38
rs762304783 Chr.11: 6478976-6478979 on Build GRCh38
rs750186715 Chr.11: 6479114-6479114 on Build GRCh38
rs201708783 Chr.11: 6478942-6478942 on Build GRCh38
rs752239238 Chr.11: 6478976-6478976 on Build GRCh38
rs1030392246 Chr.11: 6478972-6478972 on Build GRCh38
rs571970402 Chr.11: 6478965-6478965 on Build GRCh38
rs548012231 Chr.11: 6479107-6479107 on Build GRCh38
rs1017879581 Chr.11: 6479027-6479027 on Build GRCh38
rs189990514 Chr.11: 6479025-6479025 on Build GRCh38
rs777408330 Chr.11: 6479091-6479091 on Build GRCh38
rs750728743 Chr.11: 6478995-6478998 on Build GRCh38

View Full Product Details