Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63832771-63833016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMARCD2
Entrez Gene ID
6603
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2
Gene Aliases
BAF60B, CRACD2, PRO2451, Rsc6p
UniGene
Hs.250581
Gene Chromosome Location
Chr.17: 63832081-63842991 on Build GRCh38
Transcripts
NM_001098426.1 XM_005257604.2 XM_017024967.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4812887 c.1432C>T p.L478L Chr.17: 63832958-63832958 on Build GRCh38
4756300 c.1430T>C p.V477A Chr.17: 63832960-63832960 on Build GRCh38
4842418 c.1417G>T p.E473* Chr.17: 63832973-63832973 on Build GRCh38
5951007 c.1567G>A p.E523K Chr.17: 63832967-63832967 on Build GRCh38
5951008 c.1423G>A p.E475K Chr.17: 63832967-63832967 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768770584 Chr.17: 63832975-63832975 on Build GRCh38
rs575138993 Chr.17: 63832905-63832905 on Build GRCh38
rs776686551 Chr.17: 63832981-63832981 on Build GRCh38
rs79899871 Chr.17: 63832982-63832982 on Build GRCh38
rs371363888 Chr.17: 63832921-63832921 on Build GRCh38
rs181910740 Chr.17: 63832989-63832989 on Build GRCh38
rs796570333 Chr.17: 63832913-63832913 on Build GRCh38
rs995340896 Chr.17: 63832968-63832968 on Build GRCh38
rs375513592 Chr.17: 63832860-63832860 on Build GRCh38
rs144299027 Chr.17: 63832867-63832867 on Build GRCh38
rs761422612 Chr.17: 63832988-63832988 on Build GRCh38
rs778097729 Chr.17: 63832916-63832916 on Build GRCh38
rs1045326938 Chr.17: 63832879-63832879 on Build GRCh38
rs866460877 Chr.17: 63832973-63832973 on Build GRCh38
rs74421455 Chr.17: 63832927-63832927 on Build GRCh38
rs191015799 Chr.17: 63832948-63832948 on Build GRCh38
rs138256675 Chr.17: 63832846-63832846 on Build GRCh38
rs781664384 Chr.17: 63832888-63832888 on Build GRCh38
rs1053919368 Chr.17: 63832859-63832859 on Build GRCh38
rs927147384 Chr.17: 63832954-63832954 on Build GRCh38
rs2584622 Chr.17: 63832807-63832807 on Build GRCh38
rs536226182 Chr.17: 63832843-63832843 on Build GRCh38
rs935093201 Chr.17: 63832841-63832841 on Build GRCh38
rs771137408 Chr.17: 63832932-63832932 on Build GRCh38
rs775593754 Chr.17: 63832820-63832820 on Build GRCh38
rs916398127 Chr.17: 63832831-63832832 on Build GRCh38
rs775428184 Chr.17: 63832935-63832935 on Build GRCh38
rs968562547 Chr.17: 63832933-63832933 on Build GRCh38
rs746794019 Chr.17: 63832951-63832951 on Build GRCh38
rs750615020 Chr.17: 63832967-63832967 on Build GRCh38
rs142746247 Chr.17: 63832866-63832866 on Build GRCh38
rs558471495 Chr.17: 63832893-63832893 on Build GRCh38

View Full Product Details