Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35846904-35847133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM8B
Entrez Gene ID
51754
Gene Name
transmembrane protein 8B
Gene Aliases
C9orf127, NAG-5, NAG5, NGX6
UniGene
Hs.493808
Gene Chromosome Location
Chr.9: 35825817-35865515 on Build GRCh38
Transcripts
NM_001042589.2 NM_001042590.2 NM_016446.3 XM_005251483.4 XM_011517901.2 XM_011517902.2 XM_011517903.2 XM_011517904.2 XM_011517905.1 XM_011517908.2 XM_011517910.2 XM_011517911.2 XM_011517912.2 XM_011517913.2 XM_011517914.2 XM_011517915.2 XM_011517916.2 XM_011517918.2 XM_017014805.1 XM_017014806.1 XM_017014807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1108848 c.763C>T p.R255W Chr.9: 35846939-35846939 on Build GRCh38
6252738 c.819+7G>T p.? Chr.9: 35847002-35847002 on Build GRCh38
7112173 c.801C>T p.F267F Chr.9: 35846977-35846977 on Build GRCh38
1108849 c.825C>T p.C275C Chr.9: 35847001-35847001 on Build GRCh38
6804392 c.884T>C p.I295T Chr.9: 35847060-35847060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888510327 Chr.9: 35847098-35847098 on Build GRCh38
rs1048467440 Chr.9: 35847078-35847078 on Build GRCh38
rs745669140 Chr.9: 35847042-35847042 on Build GRCh38
rs753862942 Chr.9: 35846953-35846953 on Build GRCh38
rs777381951 Chr.9: 35847002-35847002 on Build GRCh38
rs768451118 Chr.9: 35847076-35847076 on Build GRCh38
rs749036375 Chr.9: 35846926-35846926 on Build GRCh38
rs771796765 Chr.9: 35847045-35847045 on Build GRCh38
rs763251368 Chr.9: 35847102-35847102 on Build GRCh38
rs749030796 Chr.9: 35846989-35846989 on Build GRCh38
rs145056456 Chr.9: 35847020-35847020 on Build GRCh38
rs758765800 Chr.9: 35846982-35846982 on Build GRCh38
rs768324747 Chr.9: 35847037-35847037 on Build GRCh38
rs1027000008 Chr.9: 35846944-35846945 on Build GRCh38
rs762169636 Chr.9: 35846963-35846963 on Build GRCh38
rs755434871 Chr.9: 35847001-35847001 on Build GRCh38
rs750845264 Chr.9: 35846974-35846974 on Build GRCh38
rs770539556 Chr.9: 35846928-35846928 on Build GRCh38
rs765536507 Chr.9: 35846964-35846964 on Build GRCh38
rs572635010 Chr.9: 35847047-35847047 on Build GRCh38
rs748722959 Chr.9: 35847014-35847014 on Build GRCh38
rs761938854 Chr.9: 35847095-35847095 on Build GRCh38
rs1025559068 Chr.9: 35847010-35847010 on Build GRCh38
rs759276138 Chr.9: 35846937-35846937 on Build GRCh38
rs181619999 Chr.9: 35846939-35846939 on Build GRCh38
rs552626381 Chr.9: 35846995-35846995 on Build GRCh38
rs760684061 Chr.9: 35846946-35846946 on Build GRCh38
rs776823788 Chr.9: 35847077-35847077 on Build GRCh38
rs764295703 Chr.9: 35846952-35846952 on Build GRCh38
rs773924693 Chr.9: 35847099-35847099 on Build GRCh38
rs778499142 Chr.9: 35847024-35847024 on Build GRCh38
rs747038251 Chr.9: 35847050-35847050 on Build GRCh38
rs774178066 Chr.9: 35846934-35846934 on Build GRCh38
rs141283034 Chr.9: 35846994-35846994 on Build GRCh38
rs775431408 Chr.9: 35846940-35846940 on Build GRCh38

View Full Product Details