Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 186398208-186398481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf27
Entrez Gene ID
54953
Gene Name
chromosome 1 open reading frame 27
Gene Aliases
ODR4, TTG1, odr-4
UniGene
Hs.371210
Gene Chromosome Location
Chr.1: 186375758-186421375 on Build GRCh38
Transcripts
NM_001164245.1 NM_001164246.1 NM_017847.5 XM_011509660.1 XM_011509661.1 XM_017001543.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5880685 c.791C>A p.S264* Chr.1: 186398323-186398323 on Build GRCh38
1290070 c.895delA p.D300fs*6 Chr.1: 186398427-186398427 on Build GRCh38
6615252 c.815T>A p.V272D Chr.1: 186398347-186398347 on Build GRCh38
2123210 c.872T>C p.I291T Chr.1: 186398404-186398404 on Build GRCh38
6873342 c.904G>A p.V302M Chr.1: 186398436-186398436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs985321941 Chr.1: 186398243-186398243 on Build GRCh38
rs190938744 Chr.1: 186398451-186398451 on Build GRCh38
rs34714819 Chr.1: 186398351-186398352 on Build GRCh38
rs374017272 Chr.1: 186398323-186398323 on Build GRCh38
rs781763599 Chr.1: 186398438-186398438 on Build GRCh38
rs752322705 Chr.1: 186398408-186398408 on Build GRCh38
rs775843214 Chr.1: 186398338-186398338 on Build GRCh38
rs747597386 Chr.1: 186398252-186398252 on Build GRCh38
rs750589667 Chr.1: 186398441-186398441 on Build GRCh38
rs762703593 Chr.1: 186398390-186398390 on Build GRCh38
rs747518060 Chr.1: 186398318-186398318 on Build GRCh38
rs371555690 Chr.1: 186398377-186398377 on Build GRCh38
rs116150040 Chr.1: 186398272-186398272 on Build GRCh38
rs200168764 Chr.1: 186398351-186398351 on Build GRCh38
rs752029378 Chr.1: 186398429-186398429 on Build GRCh38
rs773058519 Chr.1: 186398359-186398359 on Build GRCh38
rs771871368 Chr.1: 186398346-186398346 on Build GRCh38
rs757716381 Chr.1: 186398430-186398430 on Build GRCh38
rs571147643 Chr.1: 186398361-186398361 on Build GRCh38
rs374378890 Chr.1: 186398410-186398410 on Build GRCh38
rs770209712 Chr.1: 186398337-186398337 on Build GRCh38
rs778191045 Chr.1: 186398315-186398315 on Build GRCh38
rs370246660 Chr.1: 186398276-186398276 on Build GRCh38
rs758774169 Chr.1: 186398311-186398311 on Build GRCh38
rs528590496 Chr.1: 186398335-186398335 on Build GRCh38
rs764477741 Chr.1: 186398421-186398421 on Build GRCh38
rs186533770 Chr.1: 186398340-186398340 on Build GRCh38
rs896626415 Chr.1: 186398406-186398406 on Build GRCh38
rs765876918 Chr.1: 186398273-186398273 on Build GRCh38
rs754562688 Chr.1: 186398284-186398284 on Build GRCh38
rs765586118 Chr.1: 186398295-186398295 on Build GRCh38
rs759038992 Chr.1: 186398382-186398382 on Build GRCh38
rs912359352 Chr.1: 186398256-186398256 on Build GRCh38
rs753124087 Chr.1: 186398299-186398299 on Build GRCh38
rs184103285 Chr.1: 186398327-186398327 on Build GRCh38
rs368311549 Chr.1: 186398434-186398434 on Build GRCh38
rs377103269 Chr.1: 186398360-186398360 on Build GRCh38
rs776048264 Chr.1: 186398379-186398379 on Build GRCh38

View Full Product Details