Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 170997872-170998137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC2A2
Entrez Gene ID
6514
Gene Name
solute carrier family 2 member 2
Gene Aliases
GLUT2
UniGene
Hs.167584
Gene Chromosome Location
Chr.3: 170996341-171026979 on Build GRCh38
Transcripts
NM_000340.1 NM_001278658.1 NM_001278659.1 XM_011513087.2 XM_011513089.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6163950 c.1386A>G p.G462G Chr.3: 170998092-170998092 on Build GRCh38
315293 c.1463A>T p.K488I Chr.3: 170998015-170998015 on Build GRCh38
5354972 c.1505_1507delAGA p.K502delK Chr.3: 170997971-170997973 on Build GRCh38
315294 c.1425C>A p.A475A Chr.3: 170998053-170998053 on Build GRCh38
5539139 c.1506G>A p.K502K Chr.3: 170997972-170997972 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770197371 Chr.3: 170998075-170998075 on Build GRCh38
rs748401954 Chr.3: 170998072-170998072 on Build GRCh38
rs5397 Chr.3: 170998046-170998046 on Build GRCh38
rs201797691 Chr.3: 170997989-170997989 on Build GRCh38
rs766757903 Chr.3: 170998089-170998089 on Build GRCh38
rs755934523 Chr.3: 170997918-170997918 on Build GRCh38
rs374342938 Chr.3: 170998066-170998066 on Build GRCh38
rs753575081 Chr.3: 170998025-170998025 on Build GRCh38
rs776597156 Chr.3: 170997948-170997948 on Build GRCh38
rs757137603 Chr.3: 170998039-170998039 on Build GRCh38
rs759732216 Chr.3: 170998101-170998101 on Build GRCh38
rs5398 Chr.3: 170998041-170998041 on Build GRCh38
rs966895511 Chr.3: 170997962-170997962 on Build GRCh38
rs771586150 Chr.3: 170997952-170997952 on Build GRCh38
rs764176766 Chr.3: 170997895-170997895 on Build GRCh38
rs1034983128 Chr.3: 170998092-170998092 on Build GRCh38
rs769037887 Chr.3: 170998064-170998064 on Build GRCh38
rs762305192 Chr.3: 170997983-170997983 on Build GRCh38
rs140138702 Chr.3: 170998076-170998076 on Build GRCh38
rs537256236 Chr.3: 170998050-170998050 on Build GRCh38
rs758607933 Chr.3: 170997908-170997908 on Build GRCh38
rs147959014 Chr.3: 170997922-170997922 on Build GRCh38
rs752687355 Chr.3: 170997913-170997913 on Build GRCh38
rs750463981 Chr.3: 170997905-170997905 on Build GRCh38
rs776826621 Chr.3: 170997975-170997975 on Build GRCh38
rs774542648 Chr.3: 170998094-170998094 on Build GRCh38
rs374630641 Chr.3: 170997954-170997954 on Build GRCh38
rs770462591 Chr.3: 170997946-170997946 on Build GRCh38
rs141574520 Chr.3: 170997928-170997928 on Build GRCh38
rs5399 Chr.3: 170997972-170997972 on Build GRCh38
rs781215842 Chr.3: 170997911-170997911 on Build GRCh38
rs777806589 Chr.3: 170998015-170998015 on Build GRCh38
rs761189337 Chr.3: 170997969-170997969 on Build GRCh38
rs765132996 Chr.3: 170998093-170998093 on Build GRCh38
rs556023421 Chr.3: 170998052-170998052 on Build GRCh38
rs754555670 Chr.3: 170998077-170998077 on Build GRCh38
rs756099063 Chr.3: 170998011-170998011 on Build GRCh38
rs200160167 Chr.3: 170997988-170997988 on Build GRCh38
rs1043837555 Chr.3: 170997895-170997895 on Build GRCh38
rs766600474 Chr.3: 170998007-170998007 on Build GRCh38

View Full Product Details