Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128355012-128355263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC27A4
Entrez Gene ID
10999
Gene Name
solute carrier family 27 member 4
Gene Aliases
ACSVL4, FATP4, IPS
UniGene
Hs.656699
Gene Chromosome Location
Chr.9: 128340517-128361470 on Build GRCh38
Transcripts
NM_005094.3 XM_017014222.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1105554 c.1420_1421insA p.A474fs*16 Chr.9: 128355148-128355149 on Build GRCh38
5427591 c.244+2T>C p.? Chr.9: 128355192-128355192 on Build GRCh38
6253953 c.232-6G>T p.? Chr.9: 128355172-128355172 on Build GRCh38
1460226 c.1365delC p.L457fs*36 Chr.9: 128355093-128355093 on Build GRCh38
4466428 c.1434C>T p.F478F Chr.9: 128355162-128355162 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777098482 Chr.9: 128355133-128355133 on Build GRCh38
rs751179445 Chr.9: 128355054-128355054 on Build GRCh38
rs368919168 Chr.9: 128355104-128355104 on Build GRCh38
rs777188405 Chr.9: 128355221-128355221 on Build GRCh38
rs770140694 Chr.9: 128355031-128355051 on Build GRCh38
rs779347193 Chr.9: 128355096-128355096 on Build GRCh38
rs768827874 Chr.9: 128355084-128355084 on Build GRCh38
rs779152686 Chr.9: 128355172-128355172 on Build GRCh38
rs762397717 Chr.9: 128355044-128355044 on Build GRCh38
rs1030269901 Chr.9: 128355201-128355201 on Build GRCh38
rs762561274 Chr.9: 128355139-128355141 on Build GRCh38
rs750166939 Chr.9: 128355063-128355063 on Build GRCh38
rs369764019 Chr.9: 128355213-128355213 on Build GRCh38
rs771361561 Chr.9: 128355130-128355130 on Build GRCh38
rs200670378 Chr.9: 128355159-128355159 on Build GRCh38
rs777999687 Chr.9: 128355198-128355198 on Build GRCh38
rs587776383 Chr.9: 128355215-128355215 on Build GRCh38
rs761668476 Chr.9: 128355197-128355197 on Build GRCh38
rs774047091 Chr.9: 128355141-128355141 on Build GRCh38
rs763625583 Chr.9: 128355189-128355190 on Build GRCh38
rs749429635 Chr.9: 128355077-128355077 on Build GRCh38
rs147688821 Chr.9: 128355158-128355158 on Build GRCh38
rs538337739 Chr.9: 128355235-128355235 on Build GRCh38
rs921911025 Chr.9: 128355093-128355093 on Build GRCh38
rs1045311688 Chr.9: 128355241-128355241 on Build GRCh38
rs145475190 Chr.9: 128355156-128355156 on Build GRCh38
rs931994910 Chr.9: 128355103-128355103 on Build GRCh38
rs748309868 Chr.9: 128355100-128355100 on Build GRCh38
rs763783240 Chr.9: 128355045-128355045 on Build GRCh38
rs980340110 Chr.9: 128355069-128355069 on Build GRCh38
rs773860933 Chr.9: 128355195-128355199 on Build GRCh38
rs775182777 Chr.9: 128355132-128355134 on Build GRCh38
rs587776382 Chr.9: 128355168-128355168 on Build GRCh38
rs912686983 Chr.9: 128355061-128355062 on Build GRCh38
rs756038092 Chr.9: 128355074-128355074 on Build GRCh38
rs755169878 Chr.9: 128355170-128355170 on Build GRCh38
rs761056845 Chr.9: 128355109-128355109 on Build GRCh38
rs753826536 Chr.9: 128355169-128355169 on Build GRCh38
rs150834858 Chr.9: 128355138-128355138 on Build GRCh38
rs780093092 Chr.9: 128355076-128355076 on Build GRCh38
rs569113993 Chr.9: 128355195-128355195 on Build GRCh38
rs558640688 Chr.9: 128355122-128355122 on Build GRCh38
rs141763664 Chr.9: 128355134-128355134 on Build GRCh38
rs757141833 Chr.9: 128355060-128355060 on Build GRCh38
rs748188422 Chr.9: 128355183-128355183 on Build GRCh38
rs867261956 Chr.9: 128355135-128355135 on Build GRCh38
rs909296584 Chr.9: 128355136-128355136 on Build GRCh38
rs868259567 Chr.9: 128355058-128355058 on Build GRCh38
rs747379931 Chr.9: 128355206-128355206 on Build GRCh38
rs773449802 Chr.9: 128355108-128355108 on Build GRCh38
rs139251313 Chr.9: 128355145-128355145 on Build GRCh38

View Full Product Details