Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 81285457-81285730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCO1
Entrez Gene ID
53630
Gene Name
beta-carotene oxygenase 1
Gene Aliases
BCDO, BCDO1, BCMO, BCMO1, BCO
UniGene
Hs.212172
Gene Chromosome Location
Chr.16: 81238642-81291142 on Build GRCh38
Transcripts
NM_017429.2 XM_011523109.1 XM_017023286.1 XM_017023287.1 XM_017023288.1 XM_017023289.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6610922 c.1255C>T p.R419* Chr.16: 81285587-81285587 on Build GRCh38
5055127 c.1275A>C p.G425G Chr.16: 81285607-81285607 on Build GRCh38
6049407 c.1302+5T>C p.? Chr.16: 81285639-81285639 on Build GRCh38
5940577 c.1302+6G>A p.? Chr.16: 81285640-81285640 on Build GRCh38
3889042 c.1289C>T p.P430L Chr.16: 81285621-81285621 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200504527 Chr.16: 81285639-81285639 on Build GRCh38
rs776806406 Chr.16: 81285490-81285490 on Build GRCh38
rs761019097 Chr.16: 81285638-81285638 on Build GRCh38
rs778557916 Chr.16: 81285537-81285537 on Build GRCh38
rs780433756 Chr.16: 81285612-81285612 on Build GRCh38
rs770612763 Chr.16: 81285589-81285590 on Build GRCh38
rs140259439 Chr.16: 81285521-81285521 on Build GRCh38
rs546085818 Chr.16: 81285598-81285598 on Build GRCh38
rs201192971 Chr.16: 81285525-81285525 on Build GRCh38
rs558053007 Chr.16: 81285697-81285697 on Build GRCh38
rs762634213 Chr.16: 81285508-81285508 on Build GRCh38
rs889884165 Chr.16: 81285580-81285580 on Build GRCh38
rs753796192 Chr.16: 81285530-81285530 on Build GRCh38
rs761925960 Chr.16: 81285508-81285508 on Build GRCh38
rs771716619 Chr.16: 81285632-81285632 on Build GRCh38
rs564100060 Chr.16: 81285618-81285618 on Build GRCh38
rs371879531 Chr.16: 81285661-81285661 on Build GRCh38
rs759491223 Chr.16: 81285641-81285641 on Build GRCh38
rs748475777 Chr.16: 81285496-81285496 on Build GRCh38
rs765228517 Chr.16: 81285649-81285649 on Build GRCh38
rs772138416 Chr.16: 81285631-81285631 on Build GRCh38
rs189044321 Chr.16: 81285510-81285510 on Build GRCh38
rs770148276 Chr.16: 81285506-81285509 on Build GRCh38
rs534385662 Chr.16: 81285483-81285483 on Build GRCh38
rs755036694 Chr.16: 81285533-81285533 on Build GRCh38
rs375802934 Chr.16: 81285597-81285597 on Build GRCh38
rs757924558 Chr.16: 81285558-81285558 on Build GRCh38
rs769208141 Chr.16: 81285614-81285614 on Build GRCh38
rs368261768 Chr.16: 81285554-81285554 on Build GRCh38
rs371953358 Chr.16: 81285498-81285498 on Build GRCh38
rs200527924 Chr.16: 81285555-81285555 on Build GRCh38
rs930964589 Chr.16: 81285622-81285622 on Build GRCh38
rs997867961 Chr.16: 81285669-81285669 on Build GRCh38
rs777193188 Chr.16: 81285680-81285680 on Build GRCh38
rs372527154 Chr.16: 81285588-81285588 on Build GRCh38
rs373745812 Chr.16: 81285646-81285646 on Build GRCh38
rs574226536 Chr.16: 81285496-81285496 on Build GRCh38
rs757945477 Chr.16: 81285671-81285671 on Build GRCh38
rs749790044 Chr.16: 81285613-81285613 on Build GRCh38
rs150476752 Chr.16: 81285685-81285685 on Build GRCh38
rs866361020 Chr.16: 81285611-81285611 on Build GRCh38
rs763996311 Chr.16: 81285502-81285502 on Build GRCh38
rs568526377 Chr.16: 81285640-81285640 on Build GRCh38
rs752105185 Chr.16: 81285656-81285656 on Build GRCh38
rs746168176 Chr.16: 81285600-81285600 on Build GRCh38
rs975722875 Chr.16: 81285571-81285571 on Build GRCh38
rs552665284 Chr.16: 81285494-81285494 on Build GRCh38
rs772768994 Chr.16: 81285507-81285507 on Build GRCh38
rs764127079 Chr.16: 81285499-81285499 on Build GRCh38
rs770118307 Chr.16: 81285604-81285604 on Build GRCh38
rs920963812 Chr.16: 81285587-81285587 on Build GRCh38
rs965295686 Chr.16: 81285557-81285557 on Build GRCh38

View Full Product Details