Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 11186067-11186326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAS2R42
Entrez Gene ID
353164
Gene Name
taste 2 receptor member 42
Gene Aliases
T2R24, T2R42, T2R55, TAS2R55
UniGene
Hs.553716
Gene Chromosome Location
Chr.12: 11185993-11186937 on Build GRCh38
Transcripts
NM_181429.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3870769 c.690G>A p.R230R Chr.12: 11186248-11186248 on Build GRCh38
3811075 c.832T>C p.S278P Chr.12: 11186106-11186106 on Build GRCh38
5920556 c.646C>T p.L216F Chr.12: 11186292-11186292 on Build GRCh38
692020 c.621G>T p.L207F Chr.12: 11186317-11186317 on Build GRCh38
1358984 c.745delT p.S249fs*18 Chr.12: 11186193-11186193 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376310818 Chr.12: 11186244-11186244 on Build GRCh38
rs768428348 Chr.12: 11186298-11186298 on Build GRCh38
rs747210271 Chr.12: 11186096-11186096 on Build GRCh38
rs771118774 Chr.12: 11186098-11186098 on Build GRCh38
rs763547101 Chr.12: 11186129-11186129 on Build GRCh38
rs1451772 Chr.12: 11186144-11186144 on Build GRCh38
rs765031381 Chr.12: 11186220-11186220 on Build GRCh38
rs377301073 Chr.12: 11186198-11186198 on Build GRCh38
rs758299079 Chr.12: 11186157-11186157 on Build GRCh38
rs777720147 Chr.12: 11186159-11186159 on Build GRCh38
rs760244535 Chr.12: 11186143-11186143 on Build GRCh38
rs769137067 Chr.12: 11186195-11186195 on Build GRCh38
rs752423136 Chr.12: 11186232-11186232 on Build GRCh38
rs765851653 Chr.12: 11186206-11186206 on Build GRCh38
rs749886968 Chr.12: 11186142-11186142 on Build GRCh38
rs761126829 Chr.12: 11186192-11186193 on Build GRCh38
rs764564418 Chr.12: 11186249-11186250 on Build GRCh38
rs756344277 Chr.12: 11186266-11186266 on Build GRCh38
rs757401140 Chr.12: 11186251-11186251 on Build GRCh38
rs749768585 Chr.12: 11186191-11186191 on Build GRCh38
rs767828587 Chr.12: 11186163-11186164 on Build GRCh38
rs981527552 Chr.12: 11186228-11186228 on Build GRCh38
rs1020638851 Chr.12: 11186145-11186145 on Build GRCh38
rs931319812 Chr.12: 11186196-11186196 on Build GRCh38
rs201899317 Chr.12: 11186177-11186177 on Build GRCh38
rs1669413 Chr.12: 11186175-11186175 on Build GRCh38
rs375998644 Chr.12: 11186282-11186282 on Build GRCh38
rs776183247 Chr.12: 11186108-11186108 on Build GRCh38
rs780271858 Chr.12: 11186263-11186263 on Build GRCh38
rs141369006 Chr.12: 11186156-11186156 on Build GRCh38
rs915944464 Chr.12: 11186194-11186194 on Build GRCh38
rs370910300 Chr.12: 11186289-11186289 on Build GRCh38
rs914333739 Chr.12: 11186105-11186105 on Build GRCh38
rs545745141 Chr.12: 11186146-11186146 on Build GRCh38
rs770492737 Chr.12: 11186300-11186300 on Build GRCh38
rs776304375 Chr.12: 11186208-11186208 on Build GRCh38
rs778646382 Chr.12: 11186155-11186155 on Build GRCh38
rs771085574 Chr.12: 11186164-11186164 on Build GRCh38
rs754296117 Chr.12: 11186300-11186300 on Build GRCh38
rs955908428 Chr.12: 11186291-11186291 on Build GRCh38
rs781419662 Chr.12: 11186262-11186262 on Build GRCh38
rs753101364 Chr.12: 11186193-11186193 on Build GRCh38
rs1049765499 Chr.12: 11186273-11186273 on Build GRCh38
rs749621115 Chr.12: 11186287-11186287 on Build GRCh38
rs543531807 Chr.12: 11186204-11186204 on Build GRCh38
rs77975004 Chr.12: 11186200-11186200 on Build GRCh38
rs565444533 Chr.12: 11186149-11186149 on Build GRCh38
rs781617981 Chr.12: 11186172-11186172 on Build GRCh38
rs144476878 Chr.12: 11186140-11186140 on Build GRCh38
rs751433641 Chr.12: 11186245-11186245 on Build GRCh38
rs530163269 Chr.12: 11186176-11186176 on Build GRCh38
rs966399513 Chr.12: 11186150-11186150 on Build GRCh38
rs190600436 Chr.12: 11186104-11186104 on Build GRCh38
rs759746514 Chr.12: 11186089-11186092 on Build GRCh38
rs201931397 Chr.12: 11186162-11186162 on Build GRCh38
rs374668212 Chr.12: 11186116-11186116 on Build GRCh38
rs758298691 Chr.12: 11186240-11186240 on Build GRCh38

View Full Product Details