Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 114792336-114792599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFSF15
Entrez Gene ID
9966
Gene Name
tumor necrosis factor superfamily member 15
Gene Aliases
TL1, TL1A, TNLG1B, VEGI, VEGI192A
UniGene
Hs.23349
Gene Chromosome Location
Chr.9: 114784635-114806128 on Build GRCh38
Transcripts
NM_001204344.1 NM_005118.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1635991 c.278A>G p.D93G Chr.9: 114792430-114792430 on Build GRCh38
3653396 c.301G>A p.V101I Chr.9: 114792407-114792407 on Build GRCh38
3653397 c.273C>A p.D91E Chr.9: 114792435-114792435 on Build GRCh38
3926030 c.266G>A p.R89K Chr.9: 114792442-114792442 on Build GRCh38
3788153 c.273C>T p.D91D Chr.9: 114792435-114792435 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775666236 Chr.9: 114792436-114792436 on Build GRCh38
rs747024209 Chr.9: 114792445-114792445 on Build GRCh38
rs778062353 Chr.9: 114792553-114792553 on Build GRCh38
rs758406895 Chr.9: 114792381-114792381 on Build GRCh38
rs757672135 Chr.9: 114792420-114792420 on Build GRCh38
rs750983443 Chr.9: 114792383-114792383 on Build GRCh38
rs746183936 Chr.9: 114792534-114792534 on Build GRCh38
rs754690759 Chr.9: 114792408-114792408 on Build GRCh38
rs763005168 Chr.9: 114792488-114792488 on Build GRCh38
rs369310479 Chr.9: 114792476-114792476 on Build GRCh38
rs750444655 Chr.9: 114792375-114792375 on Build GRCh38
rs371139299 Chr.9: 114792547-114792547 on Build GRCh38
rs772513068 Chr.9: 114792435-114792435 on Build GRCh38
rs766186344 Chr.9: 114792489-114792489 on Build GRCh38
rs545473100 Chr.9: 114792481-114792481 on Build GRCh38
rs369835134 Chr.9: 114792515-114792515 on Build GRCh38
rs758958908 Chr.9: 114792546-114792546 on Build GRCh38
rs780469766 Chr.9: 114792542-114792542 on Build GRCh38
rs139324037 Chr.9: 114792558-114792558 on Build GRCh38
rs373564440 Chr.9: 114792533-114792533 on Build GRCh38
rs375997040 Chr.9: 114792382-114792382 on Build GRCh38
rs746375852 Chr.9: 114792434-114792434 on Build GRCh38
rs181198827 Chr.9: 114792463-114792463 on Build GRCh38
rs747530044 Chr.9: 114792545-114792545 on Build GRCh38
rs754491637 Chr.9: 114792512-114792512 on Build GRCh38
rs892706887 Chr.9: 114792514-114792514 on Build GRCh38
rs55737197 Chr.9: 114792511-114792511 on Build GRCh38
rs866972093 Chr.9: 114792433-114792433 on Build GRCh38
rs941460618 Chr.9: 114792458-114792458 on Build GRCh38
rs371815760 Chr.9: 114792366-114792366 on Build GRCh38
rs752410781 Chr.9: 114792524-114792524 on Build GRCh38
rs147641716 Chr.9: 114792432-114792432 on Build GRCh38
rs758980080 Chr.9: 114792536-114792536 on Build GRCh38
rs747809936 Chr.9: 114792419-114792419 on Build GRCh38
rs146322170 Chr.9: 114792554-114792554 on Build GRCh38
rs768751032 Chr.9: 114792446-114792446 on Build GRCh38
rs978548002 Chr.9: 114792401-114792401 on Build GRCh38
rs780747634 Chr.9: 114792413-114792413 on Build GRCh38
rs548211757 Chr.9: 114792526-114792526 on Build GRCh38

View Full Product Details