Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 990261-990508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDUA
Entrez Gene ID
3425
Gene Name
iduronidase, alpha-L-
Gene Aliases
IDA, MPS1
UniGene
Hs.89560
Gene Chromosome Location
Chr.4: 986997-1004557 on Build GRCh38
Transcripts
NM_000203.4 NR_110313.1 XM_017008163.1 NM_022042.3 NM_134425.2 NM_213613.3

Gene Symbol
SLC26A1
Entrez Gene ID
10861
Gene Name
solute carrier family 26 member 1
Gene Aliases
CAON, EDM4, SAT-1, SAT1
UniGene
Hs.658244
Gene Chromosome Location
Chr.4: 979073-993440 on Build GRCh38
Transcripts
NM_000203.4 NR_110313.1 XM_017008163.1 NM_022042.3 NM_134425.2 NM_213613.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4126861 c.607G>A p.V203M Chr.4: 990332-990332 on Build GRCh38
1431915 c.588C>T p.G196G Chr.4: 990351-990351 on Build GRCh38
6168207 c.659C>A p.S220Y Chr.4: 990280-990280 on Build GRCh38
6415592 c.589G>A p.V197I Chr.4: 990350-990350 on Build GRCh38
5890053 c.577-1G>A p.? Chr.4: 990363-990363 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748877758 Chr.4: 990350-990350 on Build GRCh38
rs776530154 Chr.4: 990333-990333 on Build GRCh38
rs779094756 Chr.4: 990374-990374 on Build GRCh38
rs554823905 Chr.4: 990342-990342 on Build GRCh38
rs764978474 Chr.4: 990344-990344 on Build GRCh38
rs572928465 Chr.4: 990343-990343 on Build GRCh38
rs535446494 Chr.4: 990455-990455 on Build GRCh38
rs779458377 Chr.4: 990320-990320 on Build GRCh38
rs746111171 Chr.4: 990279-990279 on Build GRCh38
rs575404975 Chr.4: 990327-990327 on Build GRCh38
rs765995686 Chr.4: 990298-990298 on Build GRCh38
rs1005175058 Chr.4: 990438-990438 on Build GRCh38
rs537501987 Chr.4: 990292-990292 on Build GRCh38
rs757672514 Chr.4: 990309-990309 on Build GRCh38
rs754363446 Chr.4: 990371-990371 on Build GRCh38
rs1017858062 Chr.4: 990471-990471 on Build GRCh38
rs754520907 Chr.4: 990358-990358 on Build GRCh38
rs369647722 Chr.4: 990413-990413 on Build GRCh38
rs200417386 Chr.4: 990332-990332 on Build GRCh38
rs192314032 Chr.4: 990377-990377 on Build GRCh38
rs369163740 Chr.4: 990300-990300 on Build GRCh38
rs58780485 Chr.4: 990482-990482 on Build GRCh38
rs746034907 Chr.4: 990322-990322 on Build GRCh38
rs762803524 Chr.4: 990403-990403 on Build GRCh38
rs751237413 Chr.4: 990351-990351 on Build GRCh38
rs777403763 Chr.4: 990346-990346 on Build GRCh38
rs756739580 Chr.4: 990396-990396 on Build GRCh38
rs752571486 Chr.4: 990409-990409 on Build GRCh38
rs143250139 Chr.4: 990331-990331 on Build GRCh38
rs529881853 Chr.4: 990401-990401 on Build GRCh38
rs750809401 Chr.4: 990375-990375 on Build GRCh38
rs868602875 Chr.4: 990467-990467 on Build GRCh38
rs770689130 Chr.4: 990410-990410 on Build GRCh38
rs778445363 Chr.4: 990400-990400 on Build GRCh38
rs558397842 Chr.4: 990284-990284 on Build GRCh38
rs902930279 Chr.4: 990448-990448 on Build GRCh38
rs976046653 Chr.4: 990483-990483 on Build GRCh38
rs185729198 Chr.4: 990394-990394 on Build GRCh38
rs998523485 Chr.4: 990469-990469 on Build GRCh38
rs757868510 Chr.4: 990372-990372 on Build GRCh38
rs532650407 Chr.4: 990378-990378 on Build GRCh38
rs375888392 Chr.4: 990285-990285 on Build GRCh38
rs368056565 Chr.4: 990326-990326 on Build GRCh38
rs372848776 Chr.4: 990302-990302 on Build GRCh38
rs767184160 Chr.4: 990363-990363 on Build GRCh38
rs189948598 Chr.4: 990488-990488 on Build GRCh38
rs768693510 Chr.4: 990379-990379 on Build GRCh38
rs749867797 Chr.4: 990306-990306 on Build GRCh38
rs1024506366 Chr.4: 990439-990439 on Build GRCh38

View Full Product Details