Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 50182432-50182677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC27A2
Entrez Gene ID
11001
Gene Name
solute carrier family 27 member 2
Gene Aliases
ACSVL1, FACVL1, FATP2, HsT17226, VLACS, VLCS, hFACVL1
UniGene
Hs.11729
Gene Chromosome Location
Chr.15: 50182196-50236392 on Build GRCh38
Transcripts
NM_001159629.1 NM_003645.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7417736 c.13A>G p.I5V Chr.15: 50182440-50182440 on Build GRCh38
4055422 c.197C>T p.T66M Chr.15: 50182624-50182624 on Build GRCh38
3754349 c.142A>C p.K48Q Chr.15: 50182569-50182569 on Build GRCh38
4148816 c.50T>C p.L17P Chr.15: 50182477-50182477 on Build GRCh38
7324720 c.46C>T p.P16S Chr.15: 50182473-50182473 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747324422 Chr.15: 50182599-50182599 on Build GRCh38
rs775733748 Chr.15: 50182496-50182496 on Build GRCh38
rs759285216 Chr.15: 50182623-50182623 on Build GRCh38
rs1018493941 Chr.15: 50182561-50182561 on Build GRCh38
rs772311878 Chr.15: 50182482-50182482 on Build GRCh38
rs777652108 Chr.15: 50182579-50182579 on Build GRCh38
rs941811366 Chr.15: 50182646-50182646 on Build GRCh38
rs765977908 Chr.15: 50182617-50182617 on Build GRCh38
rs760962656 Chr.15: 50182502-50182502 on Build GRCh38
rs755258247 Chr.15: 50182649-50182649 on Build GRCh38
rs779016643 Chr.15: 50182468-50182468 on Build GRCh38
rs770213875 Chr.15: 50182656-50182656 on Build GRCh38
rs749984359 Chr.15: 50182539-50182539 on Build GRCh38
rs772082726 Chr.15: 50182587-50182587 on Build GRCh38
rs147849550 Chr.15: 50182625-50182625 on Build GRCh38
rs745970555 Chr.15: 50182581-50182581 on Build GRCh38
rs149897108 Chr.15: 50182568-50182568 on Build GRCh38
rs769000298 Chr.15: 50182506-50182506 on Build GRCh38
rs62623668 Chr.15: 50182636-50182636 on Build GRCh38
rs151078854 Chr.15: 50182461-50182461 on Build GRCh38
rs775771556 Chr.15: 50182652-50182654 on Build GRCh38
rs748533115 Chr.15: 50182655-50182655 on Build GRCh38
rs780145716 Chr.15: 50182596-50182596 on Build GRCh38
rs902516928 Chr.15: 50182495-50182495 on Build GRCh38
rs762499016 Chr.15: 50182541-50182541 on Build GRCh38
rs765807138 Chr.15: 50182538-50182538 on Build GRCh38
rs776843899 Chr.15: 50182516-50182516 on Build GRCh38
rs141946591 Chr.15: 50182604-50182604 on Build GRCh38
rs757139904 Chr.15: 50182632-50182632 on Build GRCh38
rs112867154 Chr.15: 50182601-50182601 on Build GRCh38
rs866661449 Chr.15: 50182473-50182473 on Build GRCh38
rs146217804 Chr.15: 50182615-50182615 on Build GRCh38
rs372276881 Chr.15: 50182464-50182464 on Build GRCh38
rs767050184 Chr.15: 50182624-50182624 on Build GRCh38
rs745920735 Chr.15: 50182472-50182472 on Build GRCh38
rs139094380 Chr.15: 50182562-50182562 on Build GRCh38
rs751153417 Chr.15: 50182549-50182549 on Build GRCh38
rs762554045 Chr.15: 50182616-50182616 on Build GRCh38
rs1648348 Chr.15: 50182569-50182569 on Build GRCh38
rs780228780 Chr.15: 50182634-50182634 on Build GRCh38
rs749865980 Chr.15: 50182660-50182660 on Build GRCh38
rs773723428 Chr.15: 50182618-50182618 on Build GRCh38
rs765957068 Chr.15: 50182542-50182542 on Build GRCh38
rs762457946 Chr.15: 50182537-50182537 on Build GRCh38
rs753710510 Chr.15: 50182631-50182631 on Build GRCh38
rs781515719 Chr.15: 50182653-50182653 on Build GRCh38
rs756089328 Chr.15: 50182578-50182578 on Build GRCh38
rs770446882 Chr.15: 50182612-50182612 on Build GRCh38
rs371933028 Chr.15: 50182608-50182608 on Build GRCh38
rs376395646 Chr.15: 50182658-50182658 on Build GRCh38
rs762953926 Chr.15: 50182496-50182496 on Build GRCh38
rs772042307 Chr.15: 50182607-50182607 on Build GRCh38

View Full Product Details