Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 159036879-159037152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAGAP
Entrez Gene ID
117289
Gene Name
T-cell activation RhoGTPase activating protein
Gene Aliases
ARHGAP47, FKSG15, IDDM21, TAGAP1
UniGene
Hs.529984
Gene Chromosome Location
Chr.6: 159034468-159045152 on Build GRCh38
Transcripts
NM_001278733.1 NM_054114.4 NM_152133.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1288332 c.987C>A p.S329R Chr.6: 159037036-159037036 on Build GRCh38
6849032 c.1128C>A p.Y376* Chr.6: 159036895-159036895 on Build GRCh38
6173024 c.1084G>T p.V362L Chr.6: 159036939-159036939 on Build GRCh38
6836958 c.1079G>C p.S360T Chr.6: 159036944-159036944 on Build GRCh38
3920999 c.1066G>A p.E356K Chr.6: 159036957-159036957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148545546 Chr.6: 159036974-159036974 on Build GRCh38
rs781342205 Chr.6: 159037037-159037037 on Build GRCh38
rs746941697 Chr.6: 159036916-159036916 on Build GRCh38
rs140788069 Chr.6: 159037093-159037093 on Build GRCh38
rs377718195 Chr.6: 159036901-159036901 on Build GRCh38
rs751021592 Chr.6: 159037025-159037025 on Build GRCh38
rs747602006 Chr.6: 159036906-159036906 on Build GRCh38
rs746948813 Chr.6: 159036987-159036987 on Build GRCh38
rs752367843 Chr.6: 159037018-159037018 on Build GRCh38
rs759841143 Chr.6: 159036936-159036936 on Build GRCh38
rs61753626 Chr.6: 159036992-159036992 on Build GRCh38
rs775839075 Chr.6: 159036945-159036945 on Build GRCh38
rs569240044 Chr.6: 159037079-159037079 on Build GRCh38
rs774310085 Chr.6: 159037081-159037081 on Build GRCh38
rs370376182 Chr.6: 159036988-159036988 on Build GRCh38
rs146522698 Chr.6: 159037080-159037080 on Build GRCh38
rs756704870 Chr.6: 159037029-159037029 on Build GRCh38
rs745764844 Chr.6: 159037039-159037039 on Build GRCh38
rs1014706694 Chr.6: 159036930-159036930 on Build GRCh38
rs766760704 Chr.6: 159037109-159037109 on Build GRCh38
rs772135673 Chr.6: 159037092-159037092 on Build GRCh38
rs751140366 Chr.6: 159037038-159037039 on Build GRCh38
rs764199475 Chr.6: 159036954-159036954 on Build GRCh38
rs562716751 Chr.6: 159036971-159036971 on Build GRCh38
rs763861450 Chr.6: 159037129-159037129 on Build GRCh38
rs761095052 Chr.6: 159037008-159037008 on Build GRCh38
rs762336101 Chr.6: 159036967-159036967 on Build GRCh38
rs778401093 Chr.6: 159036904-159036904 on Build GRCh38
rs762234569 Chr.6: 159036999-159036999 on Build GRCh38
rs200734731 Chr.6: 159036939-159036939 on Build GRCh38
rs768645169 Chr.6: 159036997-159036997 on Build GRCh38
rs771599062 Chr.6: 159036913-159036913 on Build GRCh38
rs777150394 Chr.6: 159037023-159037023 on Build GRCh38
rs761121568 Chr.6: 159037101-159037101 on Build GRCh38
rs748319319 Chr.6: 159037083-159037083 on Build GRCh38
rs769757092 Chr.6: 159037045-159037045 on Build GRCh38
rs145511341 Chr.6: 159037117-159037117 on Build GRCh38
rs779949611 Chr.6: 159037049-159037049 on Build GRCh38
rs112777023 Chr.6: 159036984-159036984 on Build GRCh38
rs143036639 Chr.6: 159036919-159036919 on Build GRCh38
rs762524950 Chr.6: 159037118-159037118 on Build GRCh38
rs758029090 Chr.6: 159037019-159037019 on Build GRCh38
rs139309613 Chr.6: 159036990-159036990 on Build GRCh38
rs769704785 Chr.6: 159036991-159036991 on Build GRCh38
rs752343253 Chr.6: 159036979-159036979 on Build GRCh38
rs766699345 Chr.6: 159037016-159037016 on Build GRCh38
rs948142895 Chr.6: 159037128-159037128 on Build GRCh38
rs776525307 Chr.6: 159036923-159036923 on Build GRCh38
rs935456871 Chr.6: 159037108-159037108 on Build GRCh38
rs144719803 Chr.6: 159036947-159036947 on Build GRCh38
rs773091324 Chr.6: 159036915-159036915 on Build GRCh38
rs970379221 Chr.6: 159036950-159036950 on Build GRCh38
rs758731322 Chr.6: 159037007-159037007 on Build GRCh38
rs35263580 Chr.6: 159036986-159036986 on Build GRCh38
rs778595389 Chr.6: 159036976-159036976 on Build GRCh38

View Full Product Details