Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57542655-57542922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMTNL1
Entrez Gene ID
219537
Gene Name
smoothelin like 1
Gene Aliases
CHASM
UniGene
Hs.68756
Gene Chromosome Location
Chr.11: 57536841-57550274 on Build GRCh38
Transcripts
NM_001105565.2 XM_005273819.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7161031 c.82G>C p.E28Q Chr.11: 57542670-57542670 on Build GRCh38
5539456 c.169G>A p.G57R Chr.11: 57542757-57542757 on Build GRCh38
5963727 c.283A>T p.K95* Chr.11: 57542871-57542871 on Build GRCh38
5446926 c.294T>C p.S98S Chr.11: 57542882-57542882 on Build GRCh38
4512067 c.142C>T p.P48S Chr.11: 57542730-57542730 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755524899 Chr.11: 57542816-57542816 on Build GRCh38
rs778551054 Chr.11: 57542817-57542818 on Build GRCh38
rs749530352 Chr.11: 57542828-57542828 on Build GRCh38
rs748297217 Chr.11: 57542822-57542822 on Build GRCh38
rs374292312 Chr.11: 57542684-57542684 on Build GRCh38
rs758502525 Chr.11: 57542850-57542850 on Build GRCh38
rs766136377 Chr.11: 57542736-57542736 on Build GRCh38
rs774197404 Chr.11: 57542832-57542832 on Build GRCh38
rs779902212 Chr.11: 57542695-57542695 on Build GRCh38
rs772560814 Chr.11: 57542676-57542676 on Build GRCh38
rs909308997 Chr.11: 57542753-57542753 on Build GRCh38
rs759546869 Chr.11: 57542741-57542741 on Build GRCh38
rs75192176 Chr.11: 57542854-57542854 on Build GRCh38
rs781539932 Chr.11: 57542817-57542817 on Build GRCh38
rs371388504 Chr.11: 57542891-57542891 on Build GRCh38
rs770063278 Chr.11: 57542728-57542728 on Build GRCh38
rs755672277 Chr.11: 57542758-57542758 on Build GRCh38
rs568284745 Chr.11: 57542774-57542774 on Build GRCh38
rs532199537 Chr.11: 57542815-57542816 on Build GRCh38
rs753359199 Chr.11: 57542862-57542862 on Build GRCh38
rs917792654 Chr.11: 57542791-57542791 on Build GRCh38
rs535760239 Chr.11: 57542809-57542809 on Build GRCh38
rs776778790 Chr.11: 57542699-57542699 on Build GRCh38
rs771016128 Chr.11: 57542830-57542830 on Build GRCh38
rs756922713 Chr.11: 57542778-57542778 on Build GRCh38
rs780500832 Chr.11: 57542810-57542810 on Build GRCh38
rs753668039 Chr.11: 57542721-57542721 on Build GRCh38
rs754078227 Chr.11: 57542685-57542685 on Build GRCh38
rs1007084888 Chr.11: 57542901-57542901 on Build GRCh38
rs200373819 Chr.11: 57542807-57542807 on Build GRCh38
rs762772833 Chr.11: 57542735-57542735 on Build GRCh38
rs372431051 Chr.11: 57542814-57542814 on Build GRCh38
rs1024589119 Chr.11: 57542705-57542705 on Build GRCh38
rs755224098 Chr.11: 57542706-57542706 on Build GRCh38
rs773964914 Chr.11: 57542738-57542738 on Build GRCh38
rs752264875 Chr.11: 57542752-57542752 on Build GRCh38
rs747271880 Chr.11: 57542811-57542811 on Build GRCh38
rs769838214 Chr.11: 57542823-57542823 on Build GRCh38
rs767289557 Chr.11: 57542743-57542743 on Build GRCh38
rs760301217 Chr.11: 57542851-57542851 on Build GRCh38
rs80123614 Chr.11: 57542834-57542834 on Build GRCh38
rs368471316 Chr.11: 57542727-57542727 on Build GRCh38
rs535255150 Chr.11: 57542698-57542698 on Build GRCh38
rs546767884 Chr.11: 57542770-57542770 on Build GRCh38
rs778221257 Chr.11: 57542867-57542867 on Build GRCh38
rs767546747 Chr.11: 57542840-57542840 on Build GRCh38

View Full Product Details