Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101282248-101282517 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAF7L
Entrez Gene ID
54457
Gene Name
TATA-box binding protein associated factor 7 like
Gene Aliases
CT40, TAF2Q
UniGene
Hs.223806
Gene Chromosome Location
Chr.X: 101268253-101293071 on Build GRCh38
Transcripts
NM_001168474.1 NM_024885.3 XM_005262152.1 XM_006724664.1

SNP Detail

SNP ID Chromosome Location
rs911807167 Chr.X: 101282486-101282486 on Build GRCh38
rs921936685 Chr.X: 101282425-101282425 on Build GRCh38
rs749616945 Chr.X: 101282356-101282356 on Build GRCh38
rs765604805 Chr.X: 101282304-101282304 on Build GRCh38
rs768072558 Chr.X: 101282465-101282466 on Build GRCh38
rs201872370 Chr.X: 101282392-101282392 on Build GRCh38
rs772895330 Chr.X: 101282366-101282366 on Build GRCh38
rs141209944 Chr.X: 101282358-101282358 on Build GRCh38
rs370305430 Chr.X: 101282381-101282381 on Build GRCh38
rs201202100 Chr.X: 101282469-101282469 on Build GRCh38
rs773600405 Chr.X: 101282340-101282340 on Build GRCh38
rs182155185 Chr.X: 101282481-101282481 on Build GRCh38
rs763218829 Chr.X: 101282457-101282457 on Build GRCh38
rs762356562 Chr.X: 101282483-101282483 on Build GRCh38
rs759480530 Chr.X: 101282412-101282412 on Build GRCh38
rs774638270 Chr.X: 101282314-101282315 on Build GRCh38
rs746442289 Chr.X: 101282296-101282296 on Build GRCh38
rs745448615 Chr.X: 101282414-101282414 on Build GRCh38
rs965870677 Chr.X: 101282471-101282471 on Build GRCh38
rs562793474 Chr.X: 101282440-101282440 on Build GRCh38
rs770270902 Chr.X: 101282305-101282305 on Build GRCh38

View Full Product Details