Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 223394190-223394437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC185
Entrez Gene ID
164127
Gene Name
coiled-coil domain containing 185
Gene Aliases
C1orf65
UniGene
Hs.192090
Gene Chromosome Location
Chr.1: 223393373-223395470 on Build GRCh38
Transcripts
NM_152610.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6272879 c.797G>T p.R266L Chr.1: 223394272-223394272 on Build GRCh38
1198463 c.919C>T p.R307W Chr.1: 223394394-223394394 on Build GRCh38
3746811 c.913C>A p.L305M Chr.1: 223394388-223394388 on Build GRCh38
6995740 c.956G>A p.R319H Chr.1: 223394431-223394431 on Build GRCh38
319059 c.733A>T p.S245C Chr.1: 223394208-223394208 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143126549 Chr.1: 223394229-223394229 on Build GRCh38
rs761569431 Chr.1: 223394334-223394334 on Build GRCh38
rs1020729031 Chr.1: 223394266-223394266 on Build GRCh38
rs751916392 Chr.1: 223394240-223394240 on Build GRCh38
rs772895833 Chr.1: 223394402-223394402 on Build GRCh38
rs924822575 Chr.1: 223394340-223394340 on Build GRCh38
rs1054742146 Chr.1: 223394345-223394345 on Build GRCh38
rs199694908 Chr.1: 223394344-223394344 on Build GRCh38
rs768770797 Chr.1: 223394397-223394397 on Build GRCh38
rs753180931 Chr.1: 223394243-223394243 on Build GRCh38
rs774220858 Chr.1: 223394399-223394399 on Build GRCh38
rs757701072 Chr.1: 223394264-223394264 on Build GRCh38
rs573008518 Chr.1: 223394314-223394314 on Build GRCh38
rs573566361 Chr.1: 223394393-223394393 on Build GRCh38
rs374835925 Chr.1: 223394382-223394382 on Build GRCh38
rs991530032 Chr.1: 223394232-223394232 on Build GRCh38
rs200353990 Chr.1: 223394286-223394286 on Build GRCh38
rs556677048 Chr.1: 223394244-223394244 on Build GRCh38
rs762894193 Chr.1: 223394322-223394322 on Build GRCh38
rs780384449 Chr.1: 223394394-223394394 on Build GRCh38
rs746039667 Chr.1: 223394383-223394383 on Build GRCh38
rs781542528 Chr.1: 223394291-223394291 on Build GRCh38
rs962124709 Chr.1: 223394358-223394358 on Build GRCh38
rs537916148 Chr.1: 223394234-223394234 on Build GRCh38
rs922962218 Chr.1: 223394384-223394386 on Build GRCh38
rs757107911 Chr.1: 223394217-223394217 on Build GRCh38
rs750036613 Chr.1: 223394342-223394342 on Build GRCh38
rs144076109 Chr.1: 223394273-223394273 on Build GRCh38
rs774310490 Chr.1: 223394333-223394333 on Build GRCh38
rs749293228 Chr.1: 223394395-223394395 on Build GRCh38
rs779461493 Chr.1: 223394242-223394242 on Build GRCh38
rs767318649 Chr.1: 223394341-223394341 on Build GRCh38
rs543098268 Chr.1: 223394326-223394326 on Build GRCh38
rs750121015 Chr.1: 223394233-223394233 on Build GRCh38
rs775439837 Chr.1: 223394319-223394319 on Build GRCh38
rs186875984 Chr.1: 223394374-223394374 on Build GRCh38
rs182459489 Chr.1: 223394271-223394271 on Build GRCh38
rs757647945 Chr.1: 223394381-223394381 on Build GRCh38
rs761767460 Chr.1: 223394401-223394401 on Build GRCh38
rs746100656 Chr.1: 223394304-223394304 on Build GRCh38
rs757786997 Chr.1: 223394297-223394298 on Build GRCh38
rs752161726 Chr.1: 223394380-223394380 on Build GRCh38
rs779591245 Chr.1: 223394419-223394421 on Build GRCh38
rs765854757 Chr.1: 223394359-223394359 on Build GRCh38
rs751373210 Chr.1: 223394216-223394216 on Build GRCh38

View Full Product Details