Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6945718-6945979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf57
Entrez Gene ID
113246
Gene Name
chromosome 12 open reading frame 57
Gene Aliases
C10, GRCC10
UniGene
Hs.405913
Gene Chromosome Location
Chr.12: 6943438-6946003 on Build GRCh38
Transcripts
NM_001301834.1 NM_001301836.1 NM_001301837.1 NM_001301838.1 NM_138425.3 NR_126035.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
468858 c.291C>A p.S97R Chr.12: 6945832-6945832 on Build GRCh38
2100291 c.311C>A p.A104E Chr.12: 6945852-6945852 on Build GRCh38
7248061 c.306G>A p.L102L Chr.12: 6945847-6945847 on Build GRCh38
7157395 c.234C>T p.V78V Chr.12: 6945775-6945775 on Build GRCh38
2100293 c.373G>A p.A125T Chr.12: 6945914-6945914 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371237102 Chr.12: 6945881-6945881 on Build GRCh38
rs149881070 Chr.12: 6945838-6945838 on Build GRCh38
rs782145941 Chr.12: 6945804-6945804 on Build GRCh38
rs781906709 Chr.12: 6945740-6945741 on Build GRCh38
rs374751929 Chr.12: 6945914-6945914 on Build GRCh38
rs149032026 Chr.12: 6945890-6945890 on Build GRCh38
rs782414883 Chr.12: 6945805-6945805 on Build GRCh38
rs782320858 Chr.12: 6945879-6945879 on Build GRCh38
rs781896109 Chr.12: 6945948-6945948 on Build GRCh38
rs144915095 Chr.12: 6945869-6945869 on Build GRCh38
rs781915786 Chr.12: 6945809-6945809 on Build GRCh38
rs782376245 Chr.12: 6945796-6945796 on Build GRCh38
rs782454240 Chr.12: 6945756-6945756 on Build GRCh38
rs782133839 Chr.12: 6945832-6945832 on Build GRCh38
rs139068225 Chr.12: 6945798-6945798 on Build GRCh38
rs782679418 Chr.12: 6945873-6945873 on Build GRCh38
rs782745732 Chr.12: 6945827-6945827 on Build GRCh38
rs782443019 Chr.12: 6945844-6945844 on Build GRCh38
rs781800298 Chr.12: 6945746-6945746 on Build GRCh38
rs781853414 Chr.12: 6945931-6945931 on Build GRCh38
rs782086556 Chr.12: 6945920-6945920 on Build GRCh38
rs782534827 Chr.12: 6945760-6945760 on Build GRCh38
rs782165395 Chr.12: 6945900-6945900 on Build GRCh38
rs782624197 Chr.12: 6945864-6945864 on Build GRCh38
rs112297142 Chr.12: 6945768-6945768 on Build GRCh38
rs376048939 Chr.12: 6945954-6945954 on Build GRCh38
rs781813477 Chr.12: 6945908-6945908 on Build GRCh38
rs369711590 Chr.12: 6945926-6945926 on Build GRCh38
rs145396836 Chr.12: 6945897-6945897 on Build GRCh38
rs782632383 Chr.12: 6945755-6945755 on Build GRCh38
rs782060935 Chr.12: 6945810-6945810 on Build GRCh38
rs782445828 Chr.12: 6945950-6945950 on Build GRCh38
rs782577262 Chr.12: 6945951-6945951 on Build GRCh38
rs782686405 Chr.12: 6945762-6945762 on Build GRCh38
rs781955379 Chr.12: 6945789-6945789 on Build GRCh38
rs782437137 Chr.12: 6945885-6945885 on Build GRCh38
rs782400028 Chr.12: 6945891-6945891 on Build GRCh38
rs1041306428 Chr.12: 6945788-6945788 on Build GRCh38
rs782324060 Chr.12: 6945784-6945784 on Build GRCh38
rs782513111 Chr.12: 6945852-6945852 on Build GRCh38
rs782504921 Chr.12: 6945752-6945752 on Build GRCh38
rs782496684 Chr.12: 6945863-6945863 on Build GRCh38
rs782509673 Chr.12: 6945936-6945936 on Build GRCh38
rs782777793 Chr.12: 6945741-6945741 on Build GRCh38
rs781862534 Chr.12: 6945831-6945831 on Build GRCh38
rs374912364 Chr.12: 6945860-6945860 on Build GRCh38
rs781886883 Chr.12: 6945742-6945742 on Build GRCh38
rs781853174 Chr.12: 6945909-6945909 on Build GRCh38
rs781825705 Chr.12: 6945781-6945781 on Build GRCh38
rs782694267 Chr.12: 6945904-6945904 on Build GRCh38
rs375630752 Chr.12: 6945758-6945758 on Build GRCh38
rs368683747 Chr.12: 6945794-6945794 on Build GRCh38
rs140277271 Chr.12: 6945853-6945853 on Build GRCh38
rs782672649 Chr.12: 6945942-6945942 on Build GRCh38
rs782398850 Chr.12: 6945872-6945872 on Build GRCh38

View Full Product Details