Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 186833847-186834120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSWIM2
Entrez Gene ID
151112
Gene Name
zinc finger SWIM-type containing 2
Gene Aliases
MEX, ZZZ2
UniGene
Hs.375054
Gene Chromosome Location
Chr.2: 186827480-186849170 on Build GRCh38
Transcripts
NM_182521.2 XM_011510712.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7250984 c.828+1G>T p.? Chr.2: 186833945-186833945 on Build GRCh38
572519 c.810C>T p.H270H Chr.2: 186833964-186833964 on Build GRCh38
278437 c.771A>G p.L257L Chr.2: 186834003-186834003 on Build GRCh38
2711702 c.812C>T p.T271M Chr.2: 186833962-186833962 on Build GRCh38
6834413 c.828G>T p.E276D Chr.2: 186833946-186833946 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769324984 Chr.2: 186833906-186833906 on Build GRCh38
rs761098746 Chr.2: 186833931-186833931 on Build GRCh38
rs539676702 Chr.2: 186834060-186834060 on Build GRCh38
rs201278934 Chr.2: 186833900-186833900 on Build GRCh38
rs376901560 Chr.2: 186834023-186834023 on Build GRCh38
rs764450406 Chr.2: 186833938-186833938 on Build GRCh38
rs767316396 Chr.2: 186834067-186834067 on Build GRCh38
rs751530680 Chr.2: 186834065-186834065 on Build GRCh38
rs367857657 Chr.2: 186834026-186834026 on Build GRCh38
rs562211290 Chr.2: 186833890-186833890 on Build GRCh38
rs763070322 Chr.2: 186834058-186834058 on Build GRCh38
rs1011396683 Chr.2: 186834019-186834019 on Build GRCh38
rs768359552 Chr.2: 186833897-186833897 on Build GRCh38
rs994766967 Chr.2: 186833923-186833923 on Build GRCh38
rs949725327 Chr.2: 186833881-186833881 on Build GRCh38
rs139317820 Chr.2: 186834069-186834069 on Build GRCh38
rs543910936 Chr.2: 186833879-186833879 on Build GRCh38
rs765496222 Chr.2: 186834048-186834048 on Build GRCh38
rs999494804 Chr.2: 186834008-186834008 on Build GRCh38
rs769160898 Chr.2: 186834028-186834028 on Build GRCh38
rs775289218 Chr.2: 186833998-186833998 on Build GRCh38
rs780926419 Chr.2: 186833982-186833982 on Build GRCh38
rs748815970 Chr.2: 186834017-186834017 on Build GRCh38
rs560447554 Chr.2: 186833909-186833909 on Build GRCh38
rs755735033 Chr.2: 186834001-186834001 on Build GRCh38
rs549159223 Chr.2: 186833971-186833971 on Build GRCh38
rs772626949 Chr.2: 186833924-186833924 on Build GRCh38
rs371673588 Chr.2: 186834040-186834040 on Build GRCh38
rs754514742 Chr.2: 186834092-186834092 on Build GRCh38
rs750447167 Chr.2: 186833961-186833961 on Build GRCh38
rs6434197 Chr.2: 186833980-186833980 on Build GRCh38
rs375968688 Chr.2: 186834055-186834055 on Build GRCh38
rs901182876 Chr.2: 186834002-186834002 on Build GRCh38
rs752424883 Chr.2: 186834080-186834080 on Build GRCh38
rs34506733 Chr.2: 186834090-186834091 on Build GRCh38
rs879177995 Chr.2: 186834062-186834062 on Build GRCh38
rs77894289 Chr.2: 186833905-186833905 on Build GRCh38
rs776539237 Chr.2: 186834073-186834073 on Build GRCh38
rs771795921 Chr.2: 186833952-186833952 on Build GRCh38
rs775773317 Chr.2: 186833929-186833929 on Build GRCh38
rs754140555 Chr.2: 186833947-186833947 on Build GRCh38
rs200253183 Chr.2: 186833950-186833950 on Build GRCh38
rs759976411 Chr.2: 186833917-186833917 on Build GRCh38
rs543166048 Chr.2: 186834060-186834061 on Build GRCh38
rs796134054 Chr.2: 186834061-186834061 on Build GRCh38
rs751408387 Chr.2: 186833979-186833979 on Build GRCh38
rs188840579 Chr.2: 186834016-186834016 on Build GRCh38
rs747820640 Chr.2: 186833992-186833992 on Build GRCh38
rs372274270 Chr.2: 186833962-186833962 on Build GRCh38
rs7559533 Chr.2: 186834024-186834024 on Build GRCh38

View Full Product Details