Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 2229037-2229310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SKI
Entrez Gene ID
6497
Gene Name
SKI proto-oncogene
Gene Aliases
SGS, SKV
UniGene
Hs.656507
Gene Chromosome Location
Chr.1: 2228695-2310213 on Build GRCh38
Transcripts
NM_003036.3 XM_005244775.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4494721 c.438C>T p.L146L Chr.1: 2229204-2229204 on Build GRCh38
5954337 c.404C>T p.S135L Chr.1: 2229170-2229170 on Build GRCh38
1931357 c.339C>T p.F113F Chr.1: 2229105-2229105 on Build GRCh38
1931366 c.394C>T p.R132C Chr.1: 2229160-2229160 on Build GRCh38
6374174 c.379C>A p.L127I Chr.1: 2229145-2229145 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886039149 Chr.1: 2229077-2229077 on Build GRCh38
rs149898447 Chr.1: 2229222-2229222 on Build GRCh38
rs867726673 Chr.1: 2229229-2229229 on Build GRCh38
rs373291824 Chr.1: 2229255-2229255 on Build GRCh38
rs748004059 Chr.1: 2229159-2229159 on Build GRCh38
rs201144990 Chr.1: 2229090-2229090 on Build GRCh38
rs564817940 Chr.1: 2229084-2229084 on Build GRCh38
rs762205602 Chr.1: 2229188-2229188 on Build GRCh38
rs371061756 Chr.1: 2229204-2229204 on Build GRCh38
rs952491836 Chr.1: 2229246-2229246 on Build GRCh38
rs387907303 Chr.1: 2229113-2229113 on Build GRCh38
rs368195821 Chr.1: 2229078-2229078 on Build GRCh38
rs869025525 Chr.1: 2229118-2229118 on Build GRCh38
rs752031916 Chr.1: 2229102-2229102 on Build GRCh38
rs763083721 Chr.1: 2229177-2229177 on Build GRCh38
rs937379586 Chr.1: 2229165-2229165 on Build GRCh38
rs369090536 Chr.1: 2229144-2229144 on Build GRCh38
rs1026586604 Chr.1: 2229231-2229231 on Build GRCh38
rs559020511 Chr.1: 2229230-2229230 on Build GRCh38
rs878854599 Chr.1: 2229253-2229253 on Build GRCh38
rs767973122 Chr.1: 2229189-2229189 on Build GRCh38
rs182513685 Chr.1: 2229282-2229282 on Build GRCh38
rs762117658 Chr.1: 2229277-2229277 on Build GRCh38
rs771747350 Chr.1: 2229288-2229288 on Build GRCh38
rs774588048 Chr.1: 2229270-2229270 on Build GRCh38
rs1013962872 Chr.1: 2229186-2229186 on Build GRCh38
rs375024753 Chr.1: 2229126-2229126 on Build GRCh38
rs150306072 Chr.1: 2229162-2229162 on Build GRCh38
rs763316588 Chr.1: 2229093-2229093 on Build GRCh38
rs569206617 Chr.1: 2229237-2229237 on Build GRCh38
rs765800099 Chr.1: 2229207-2229207 on Build GRCh38
rs777792553 Chr.1: 2229234-2229234 on Build GRCh38
rs144874401 Chr.1: 2229183-2229183 on Build GRCh38
rs768420010 Chr.1: 2229150-2229150 on Build GRCh38
rs546957439 Chr.1: 2229213-2229213 on Build GRCh38
rs764373577 Chr.1: 2229096-2229096 on Build GRCh38
rs115746142 Chr.1: 2229060-2229060 on Build GRCh38
rs772192813 Chr.1: 2229069-2229069 on Build GRCh38
rs770970008 Chr.1: 2229247-2229247 on Build GRCh38
rs769774481 Chr.1: 2229171-2229171 on Build GRCh38
rs863223723 Chr.1: 2229112-2229112 on Build GRCh38
rs768733290 Chr.1: 2229264-2229264 on Build GRCh38
rs538703800 Chr.1: 2229216-2229216 on Build GRCh38
rs753444459 Chr.1: 2229208-2229208 on Build GRCh38
rs773310002 Chr.1: 2229168-2229168 on Build GRCh38
rs863223724 Chr.1: 2229265-2229266 on Build GRCh38
rs749438586 Chr.1: 2229261-2229261 on Build GRCh38
rs566906261 Chr.1: 2229219-2229219 on Build GRCh38
rs775713153 Chr.1: 2229172-2229172 on Build GRCh38
rs979256214 Chr.1: 2229086-2229086 on Build GRCh38
rs757751828 Chr.1: 2229107-2229107 on Build GRCh38
rs776829569 Chr.1: 2229075-2229075 on Build GRCh38
rs567637491 Chr.1: 2229127-2229127 on Build GRCh38
rs113737327 Chr.1: 2229054-2229054 on Build GRCh38
rs869312901 Chr.1: 2229115-2229115 on Build GRCh38
rs750951866 Chr.1: 2229195-2229195 on Build GRCh38
rs371497152 Chr.1: 2229147-2229147 on Build GRCh38
rs781703488 Chr.1: 2229117-2229117 on Build GRCh38

View Full Product Details