Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 183248627-183248898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WWC2
Entrez Gene ID
80014
Gene Name
WW and C2 domain containing 2
Gene Aliases
BOMB
UniGene
Hs.333179
Gene Chromosome Location
Chr.4: 183098677-183320777 on Build GRCh38
Transcripts
NM_024949.5 XM_011532269.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7352325 c.879+1G>A p.? Chr.4: 183248861-183248861 on Build GRCh38
5766778 c.580G>A p.G194R Chr.4: 183248855-183248855 on Build GRCh38
7352324 c.879+1G>A p.? Chr.4: 183248861-183248861 on Build GRCh38
3696557 c.779G>A p.G260D Chr.4: 183248760-183248760 on Build GRCh38
7189004 c.825A>G p.L275L Chr.4: 183248806-183248806 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759054918 Chr.4: 183248777-183248777 on Build GRCh38
rs761338914 Chr.4: 183248693-183248693 on Build GRCh38
rs751652135 Chr.4: 183248839-183248839 on Build GRCh38
rs373959652 Chr.4: 183248791-183248791 on Build GRCh38
rs771560744 Chr.4: 183248762-183248762 on Build GRCh38
rs773710257 Chr.4: 183248679-183248680 on Build GRCh38
rs777272737 Chr.4: 183248798-183248798 on Build GRCh38
rs767211039 Chr.4: 183248676-183248676 on Build GRCh38
rs765212126 Chr.4: 183248735-183248735 on Build GRCh38
rs755002850 Chr.4: 183248854-183248854 on Build GRCh38
rs73870396 Chr.4: 183248659-183248659 on Build GRCh38
rs577462909 Chr.4: 183248678-183248678 on Build GRCh38
rs752550316 Chr.4: 183248690-183248690 on Build GRCh38
rs765096616 Chr.4: 183248833-183248833 on Build GRCh38
rs781334849 Chr.4: 183248748-183248748 on Build GRCh38
rs767296574 Chr.4: 183248784-183248784 on Build GRCh38
rs780169085 Chr.4: 183248734-183248734 on Build GRCh38
rs750398316 Chr.4: 183248739-183248739 on Build GRCh38
rs73870397 Chr.4: 183248675-183248675 on Build GRCh38
rs907357 Chr.4: 183248872-183248872 on Build GRCh38
rs761727757 Chr.4: 183248824-183248824 on Build GRCh38
rs764776213 Chr.4: 183248712-183248713 on Build GRCh38
rs757969705 Chr.4: 183248764-183248764 on Build GRCh38
rs969313587 Chr.4: 183248687-183248687 on Build GRCh38
rs146332278 Chr.4: 183248671-183248671 on Build GRCh38
rs764121683 Chr.4: 183248698-183248698 on Build GRCh38
rs763847726 Chr.4: 183248812-183248812 on Build GRCh38
rs752030516 Chr.4: 183248761-183248762 on Build GRCh38
rs146190351 Chr.4: 183248823-183248823 on Build GRCh38
rs753756289 Chr.4: 183248701-183248701 on Build GRCh38
rs574720107 Chr.4: 183248855-183248855 on Build GRCh38
rs760341299 Chr.4: 183248803-183248803 on Build GRCh38
rs774002337 Chr.4: 183248774-183248774 on Build GRCh38
rs755201324 Chr.4: 183248736-183248736 on Build GRCh38
rs756344952 Chr.4: 183248864-183248864 on Build GRCh38
rs752904297 Chr.4: 183248861-183248861 on Build GRCh38
rs757200621 Chr.4: 183248720-183248720 on Build GRCh38
rs538336229 Chr.4: 183248760-183248760 on Build GRCh38
rs770016260 Chr.4: 183248757-183248757 on Build GRCh38

View Full Product Details