Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 34541115-34541348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPDEF
Entrez Gene ID
25803
Gene Name
SAM pointed domain containing ETS transcription factor
Gene Aliases
PDEF, bA375E1.3
UniGene
Hs.485158
Gene Chromosome Location
Chr.6: 34537802-34556333 on Build GRCh38
Transcripts
NM_001252294.1 NM_012391.2 XM_005248988.4 XM_011514457.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4494773 c.439C>T p.P147S Chr.6: 34541179-34541179 on Build GRCh38
3928407 c.478C>T p.L160L Chr.6: 34541140-34541140 on Build GRCh38
3350908 c.449G>A p.W150* Chr.6: 34541169-34541169 on Build GRCh38
6189198 c.490C>T p.H164Y Chr.6: 34541128-34541128 on Build GRCh38
42933 c.440C>G p.P147R Chr.6: 34541178-34541178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535848938 Chr.6: 34541192-34541192 on Build GRCh38
rs183969187 Chr.6: 34541244-34541244 on Build GRCh38
rs906776060 Chr.6: 34541327-34541327 on Build GRCh38
rs779509332 Chr.6: 34541223-34541224 on Build GRCh38
rs943957712 Chr.6: 34541243-34541243 on Build GRCh38
rs936673503 Chr.6: 34541272-34541273 on Build GRCh38
rs2233642 Chr.6: 34541176-34541176 on Build GRCh38
rs201218077 Chr.6: 34541212-34541212 on Build GRCh38
rs779713831 Chr.6: 34541181-34541181 on Build GRCh38
rs973822994 Chr.6: 34541153-34541153 on Build GRCh38
rs561444179 Chr.6: 34541275-34541275 on Build GRCh38
rs773106130 Chr.6: 34541221-34541221 on Build GRCh38
rs373970589 Chr.6: 34541236-34541236 on Build GRCh38
rs978153997 Chr.6: 34541223-34541223 on Build GRCh38
rs528994264 Chr.6: 34541301-34541301 on Build GRCh38
rs752115907 Chr.6: 34541159-34541159 on Build GRCh38
rs546199525 Chr.6: 34541228-34541228 on Build GRCh38
rs781665049 Chr.6: 34541161-34541161 on Build GRCh38
rs564665171 Chr.6: 34541241-34541241 on Build GRCh38
rs945558474 Chr.6: 34541299-34541299 on Build GRCh38
rs748182166 Chr.6: 34541199-34541199 on Build GRCh38
rs975395566 Chr.6: 34541247-34541247 on Build GRCh38
rs540452666 Chr.6: 34541274-34541274 on Build GRCh38
rs748746776 Chr.6: 34541185-34541185 on Build GRCh38
rs376752129 Chr.6: 34541150-34541150 on Build GRCh38
rs921236248 Chr.6: 34541270-34541270 on Build GRCh38
rs1054219770 Chr.6: 34541293-34541293 on Build GRCh38
rs932574141 Chr.6: 34541328-34541328 on Build GRCh38
rs757560677 Chr.6: 34541160-34541160 on Build GRCh38
rs112447566 Chr.6: 34541324-34541324 on Build GRCh38
rs750022965 Chr.6: 34541165-34541165 on Build GRCh38
rs1053835262 Chr.6: 34541282-34541282 on Build GRCh38
rs376417530 Chr.6: 34541195-34541195 on Build GRCh38

View Full Product Details