Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65038391-65038650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL2-SNX15
Entrez Gene ID
100528018
Gene Name
ARL2-SNX15 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.80132
Gene Chromosome Location
Chr.11: 65014113-65040572 on Build GRCh38
Transcripts
NR_037650.1 NM_013306.4 NM_147777.3

Gene Symbol
SNX15
Entrez Gene ID
29907
Gene Name
sorting nexin 15
Gene Aliases
HSAF001435
UniGene
Hs.80132
Gene Chromosome Location
Chr.11: 65027408-65040572 on Build GRCh38
Transcripts
NR_037650.1 NM_013306.4 NM_147777.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7320565 c.703G>A p.A235T Chr.11: 65038610-65038610 on Build GRCh38
1227042 c.707C>T p.T236M Chr.11: 65038614-65038614 on Build GRCh38
4450542 c.665-1G>A p.? Chr.11: 65038571-65038571 on Build GRCh38
1227039 c.670G>A p.A224T Chr.11: 65038577-65038577 on Build GRCh38
689626 c.688C>G p.H230D Chr.11: 65038595-65038595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576985248 Chr.11: 65038508-65038508 on Build GRCh38
rs990030341 Chr.11: 65038562-65038562 on Build GRCh38
rs902051261 Chr.11: 65038631-65038631 on Build GRCh38
rs931538044 Chr.11: 65038454-65038454 on Build GRCh38
rs138353324 Chr.11: 65038577-65038577 on Build GRCh38
rs953185961 Chr.11: 65038554-65038554 on Build GRCh38
rs879160495 Chr.11: 65038607-65038607 on Build GRCh38
rs200619885 Chr.11: 65038549-65038549 on Build GRCh38
rs894608035 Chr.11: 65038453-65038453 on Build GRCh38
rs572444822 Chr.11: 65038540-65038540 on Build GRCh38
rs1055477255 Chr.11: 65038446-65038446 on Build GRCh38
rs369228727 Chr.11: 65038587-65038587 on Build GRCh38
rs763563527 Chr.11: 65038566-65038566 on Build GRCh38
rs568401339 Chr.11: 65038430-65038430 on Build GRCh38
rs955824035 Chr.11: 65038423-65038423 on Build GRCh38
rs557267530 Chr.11: 65038482-65038482 on Build GRCh38
rs199551088 Chr.11: 65038539-65038539 on Build GRCh38
rs765727483 Chr.11: 65038551-65038551 on Build GRCh38
rs537323925 Chr.11: 65038469-65038469 on Build GRCh38
rs919904669 Chr.11: 65038613-65038613 on Build GRCh38
rs778568224 Chr.11: 65038576-65038576 on Build GRCh38
rs748021249 Chr.11: 65038599-65038599 on Build GRCh38
rs199545875 Chr.11: 65038596-65038596 on Build GRCh38
rs370680178 Chr.11: 65038548-65038548 on Build GRCh38
rs577473650 Chr.11: 65038459-65038459 on Build GRCh38
rs746200451 Chr.11: 65038602-65038602 on Build GRCh38
rs771832927 Chr.11: 65038601-65038601 on Build GRCh38
rs539220338 Chr.11: 65038517-65038517 on Build GRCh38
rs377428612 Chr.11: 65038598-65038598 on Build GRCh38
rs267603110 Chr.11: 65038589-65038589 on Build GRCh38
rs917100648 Chr.11: 65038448-65038448 on Build GRCh38
rs769192419 Chr.11: 65038585-65038586 on Build GRCh38
rs377480359 Chr.11: 65038544-65038544 on Build GRCh38

View Full Product Details