Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42511742-42512003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNAP23
Entrez Gene ID
8773
Gene Name
synaptosome associated protein 23
Gene Aliases
HsT17016, SNAP-23, SNAP23A, SNAP23B
UniGene
Hs.511149
Gene Chromosome Location
Chr.15: 42491244-42533061 on Build GRCh38
Transcripts
NM_003825.3 NM_130798.2 XM_006720725.2 XM_017022692.1 XM_017022693.1 XM_017022694.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6198911 c.13T>A p.S5T Chr.15: 42511859-42511859 on Build GRCh38
1266324 c.28C>T p.Q10* Chr.15: 42511874-42511874 on Build GRCh38
4578252 c.55G>A p.E19K Chr.15: 42511901-42511901 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs181598526 Chr.15: 42511850-42511850 on Build GRCh38
rs762598444 Chr.15: 42511811-42511811 on Build GRCh38
rs377114237 Chr.15: 42511844-42511844 on Build GRCh38
rs781067904 Chr.15: 42511875-42511875 on Build GRCh38
rs772765873 Chr.15: 42511808-42511808 on Build GRCh38
rs144588092 Chr.15: 42511967-42511967 on Build GRCh38
rs752740298 Chr.15: 42511886-42511886 on Build GRCh38
rs781299181 Chr.15: 42511798-42511798 on Build GRCh38
rs1047979178 Chr.15: 42511792-42511792 on Build GRCh38
rs777189318 Chr.15: 42511901-42511901 on Build GRCh38
rs563982657 Chr.15: 42511935-42511935 on Build GRCh38
rs140830704 Chr.15: 42511950-42511950 on Build GRCh38
rs746787924 Chr.15: 42511882-42511882 on Build GRCh38
rs1056600464 Chr.15: 42511834-42511834 on Build GRCh38
rs750622440 Chr.15: 42511869-42511869 on Build GRCh38
rs748811679 Chr.15: 42511916-42511916 on Build GRCh38
rs1007839662 Chr.15: 42511976-42511976 on Build GRCh38
rs111858985 Chr.15: 42511894-42511894 on Build GRCh38
rs765473822 Chr.15: 42511855-42511855 on Build GRCh38
rs1012713240 Chr.15: 42511876-42511876 on Build GRCh38
rs907903256 Chr.15: 42511794-42511794 on Build GRCh38
rs895224205 Chr.15: 42511848-42511848 on Build GRCh38
rs529657952 Chr.15: 42511936-42511936 on Build GRCh38
rs770587678 Chr.15: 42511822-42511822 on Build GRCh38
rs369461741 Chr.15: 42511872-42511872 on Build GRCh38
rs147883842 Chr.15: 42511979-42511979 on Build GRCh38
rs903663048 Chr.15: 42511917-42511917 on Build GRCh38
rs774749400 Chr.15: 42511823-42511823 on Build GRCh38
rs761979101 Chr.15: 42511852-42511852 on Build GRCh38
rs769191186 Chr.15: 42511805-42511805 on Build GRCh38
rs200992797 Chr.15: 42511826-42511826 on Build GRCh38
rs772389017 Chr.15: 42511833-42511833 on Build GRCh38
rs776754560 Chr.15: 42511842-42511843 on Build GRCh38
rs564634418 Chr.15: 42511846-42511846 on Build GRCh38

View Full Product Details