Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122206974-122207219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GNT4
Entrez Gene ID
79369
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 4
Gene Aliases
B3GN-T4, beta3Gn-T4
UniGene
Hs.363315
Gene Chromosome Location
Chr.12: 122203681-122208135 on Build GRCh38
Transcripts
NM_030765.2 XM_011538732.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1184318 c.725T>C p.L242P Chr.12: 122206976-122206976 on Build GRCh38
6609321 c.904C>T p.P302S Chr.12: 122207155-122207155 on Build GRCh38
202590 c.869G>A p.R290H Chr.12: 122207120-122207120 on Build GRCh38
7251856 c.874C>A p.Q292K Chr.12: 122207125-122207125 on Build GRCh38
5559219 c.951G>T p.L317L Chr.12: 122207202-122207202 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749195459 Chr.12: 122207134-122207134 on Build GRCh38
rs765215253 Chr.12: 122207011-122207011 on Build GRCh38
rs756818381 Chr.12: 122207167-122207167 on Build GRCh38
rs947010499 Chr.12: 122207058-122207058 on Build GRCh38
rs760975394 Chr.12: 122207200-122207200 on Build GRCh38
rs753666719 Chr.12: 122207189-122207191 on Build GRCh38
rs529236293 Chr.12: 122207045-122207045 on Build GRCh38
rs572271254 Chr.12: 122207047-122207047 on Build GRCh38
rs766818650 Chr.12: 122207156-122207156 on Build GRCh38
rs773600437 Chr.12: 122207145-122207145 on Build GRCh38
rs572008767 Chr.12: 122207196-122207196 on Build GRCh38
rs767745310 Chr.12: 122207031-122207031 on Build GRCh38
rs761968464 Chr.12: 122207136-122207136 on Build GRCh38
rs956743677 Chr.12: 122207003-122207003 on Build GRCh38
rs773463607 Chr.12: 122207197-122207197 on Build GRCh38
rs148329384 Chr.12: 122207063-122207063 on Build GRCh38
rs750759224 Chr.12: 122207091-122207091 on Build GRCh38
rs755579614 Chr.12: 122207112-122207112 on Build GRCh38
rs768708305 Chr.12: 122207135-122207135 on Build GRCh38
rs769603688 Chr.12: 122207128-122207128 on Build GRCh38
rs750059163 Chr.12: 122207172-122207172 on Build GRCh38
rs759462178 Chr.12: 122207009-122207009 on Build GRCh38
rs751755393 Chr.12: 122207025-122207025 on Build GRCh38
rs756552864 Chr.12: 122207093-122207093 on Build GRCh38
rs140179789 Chr.12: 122207129-122207129 on Build GRCh38
rs200567354 Chr.12: 122207190-122207190 on Build GRCh38
rs767839496 Chr.12: 122207082-122207082 on Build GRCh38
rs570110830 Chr.12: 122207119-122207119 on Build GRCh38
rs752212574 Chr.12: 122207189-122207189 on Build GRCh38
rs775486046 Chr.12: 122207012-122207012 on Build GRCh38
rs754320135 Chr.12: 122207161-122207161 on Build GRCh38
rs549584174 Chr.12: 122207181-122207181 on Build GRCh38
rs774634761 Chr.12: 122207057-122207057 on Build GRCh38
rs780663730 Chr.12: 122207169-122207169 on Build GRCh38
rs763020841 Chr.12: 122207021-122207021 on Build GRCh38
rs868683322 Chr.12: 122207076-122207076 on Build GRCh38
rs957029254 Chr.12: 122207186-122207186 on Build GRCh38
rs769648867 Chr.12: 122207060-122207060 on Build GRCh38
rs764263985 Chr.12: 122207023-122207023 on Build GRCh38
rs4758675 Chr.12: 122207191-122207191 on Build GRCh38
rs747595280 Chr.12: 122207043-122207043 on Build GRCh38
rs764060683 Chr.12: 122207064-122207064 on Build GRCh38
rs750965599 Chr.12: 122207166-122207166 on Build GRCh38
rs777391992 Chr.12: 122207046-122207046 on Build GRCh38
rs1021497816 Chr.12: 122207019-122207020 on Build GRCh38
rs939477759 Chr.12: 122207078-122207078 on Build GRCh38
rs141506774 Chr.12: 122207117-122207117 on Build GRCh38
rs745673381 Chr.12: 122207055-122207055 on Build GRCh38
rs59453864 Chr.12: 122206994-122206994 on Build GRCh38
rs746769802 Chr.12: 122206993-122206993 on Build GRCh38
rs774533386 Chr.12: 122207070-122207070 on Build GRCh38
rs761027036 Chr.12: 122207148-122207148 on Build GRCh38
rs373036509 Chr.12: 122207052-122207052 on Build GRCh38
rs772119867 Chr.12: 122207139-122207139 on Build GRCh38
rs750868441 Chr.12: 122207032-122207032 on Build GRCh38
rs772851416 Chr.12: 122207160-122207162 on Build GRCh38
rs374005009 Chr.12: 122207094-122207094 on Build GRCh38
rs988575747 Chr.12: 122207042-122207042 on Build GRCh38
rs997729109 Chr.12: 122207168-122207168 on Build GRCh38
rs780720644 Chr.12: 122207039-122207039 on Build GRCh38
rs938324322 Chr.12: 122207164-122207164 on Build GRCh38
rs754373696 Chr.12: 122207111-122207111 on Build GRCh38
rs144265607 Chr.12: 122207053-122207053 on Build GRCh38
rs757550904 Chr.12: 122207026-122207026 on Build GRCh38
rs1056237275 Chr.12: 122207081-122207081 on Build GRCh38
rs766942993 Chr.12: 122207099-122207099 on Build GRCh38
rs926801135 Chr.12: 122207071-122207071 on Build GRCh38
rs147517845 Chr.12: 122207120-122207120 on Build GRCh38
rs1002552024 Chr.12: 122207000-122207000 on Build GRCh38
rs765785037 Chr.12: 122207165-122207165 on Build GRCh38
rs756677704 Chr.12: 122207033-122207033 on Build GRCh38
rs910089724 Chr.12: 122207084-122207084 on Build GRCh38
rs897543888 Chr.12: 122207113-122207113 on Build GRCh38
rs1057256645 Chr.12: 122207005-122207005 on Build GRCh38
rs7965804 Chr.12: 122207192-122207192 on Build GRCh38
rs759837170 Chr.12: 122207147-122207148 on Build GRCh38
rs755805957 Chr.12: 122207176-122207176 on Build GRCh38
rs761938290 Chr.12: 122207075-122207075 on Build GRCh38

View Full Product Details