Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1990610-1990845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD2
Entrez Gene ID
55643
Gene Name
BTB domain containing 2
Gene Aliases
-
UniGene
Hs.465543
Gene Chromosome Location
Chr.19: 1985438-2015703 on Build GRCh38
Transcripts
NM_017797.3 XM_005259593.1 XM_011528127.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
993435 c.754G>A p.A252T Chr.19: 1990753-1990753 on Build GRCh38
6273294 c.774C>A p.F258L Chr.19: 1990733-1990733 on Build GRCh38
993436 c.753C>T p.D251D Chr.19: 1990754-1990754 on Build GRCh38
5982387 c.696C>T p.F232F Chr.19: 1990811-1990811 on Build GRCh38
993437 c.688C>T p.R230* Chr.19: 1990819-1990819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746382666 Chr.19: 1990684-1990684 on Build GRCh38
rs1032674213 Chr.19: 1990688-1990688 on Build GRCh38
rs766195972 Chr.19: 1990730-1990730 on Build GRCh38
rs933745248 Chr.19: 1990660-1990660 on Build GRCh38
rs752764052 Chr.19: 1990670-1990670 on Build GRCh38
rs747657912 Chr.19: 1990789-1990789 on Build GRCh38
rs779379687 Chr.19: 1990681-1990681 on Build GRCh38
rs1031843350 Chr.19: 1990744-1990744 on Build GRCh38
rs781156794 Chr.19: 1990669-1990669 on Build GRCh38
rs757504334 Chr.19: 1990767-1990767 on Build GRCh38
rs80054150 Chr.19: 1990784-1990784 on Build GRCh38
rs755531881 Chr.19: 1990687-1990690 on Build GRCh38
rs866719892 Chr.19: 1990632-1990632 on Build GRCh38
rs751323075 Chr.19: 1990736-1990736 on Build GRCh38
rs772672296 Chr.19: 1990803-1990803 on Build GRCh38
rs991030622 Chr.19: 1990716-1990716 on Build GRCh38
rs889481333 Chr.19: 1990678-1990678 on Build GRCh38
rs761377857 Chr.19: 1990697-1990697 on Build GRCh38
rs756206197 Chr.19: 1990753-1990753 on Build GRCh38
rs376863189 Chr.19: 1990672-1990672 on Build GRCh38
rs1036879109 Chr.19: 1990641-1990641 on Build GRCh38
rs113462365 Chr.19: 1990817-1990817 on Build GRCh38
rs772573538 Chr.19: 1990695-1990695 on Build GRCh38
rs748965481 Chr.19: 1990811-1990811 on Build GRCh38
rs749750917 Chr.19: 1990671-1990671 on Build GRCh38
rs1037288334 Chr.19: 1990820-1990820 on Build GRCh38
rs759408613 Chr.19: 1990742-1990742 on Build GRCh38
rs758890370 Chr.19: 1990772-1990772 on Build GRCh38
rs749730869 Chr.19: 1990675-1990675 on Build GRCh38
rs769261249 Chr.19: 1990802-1990802 on Build GRCh38
rs146922665 Chr.19: 1990754-1990754 on Build GRCh38
rs8102506 Chr.19: 1990826-1990826 on Build GRCh38
rs776004357 Chr.19: 1990696-1990696 on Build GRCh38
rs376436365 Chr.19: 1990712-1990712 on Build GRCh38
rs137893930 Chr.19: 1990760-1990760 on Build GRCh38
rs762778997 Chr.19: 1990724-1990724 on Build GRCh38
rs752787818 Chr.19: 1990751-1990751 on Build GRCh38
rs371735283 Chr.19: 1990700-1990700 on Build GRCh38
rs150224120 Chr.19: 1990667-1990677 on Build GRCh38
rs143448816 Chr.19: 1990770-1990770 on Build GRCh38
rs4806826 Chr.19: 1990680-1990680 on Build GRCh38
rs755704223 Chr.19: 1990659-1990659 on Build GRCh38
rs1022316750 Chr.19: 1990683-1990683 on Build GRCh38
rs778138953 Chr.19: 1990673-1990673 on Build GRCh38
rs746177049 Chr.19: 1990771-1990771 on Build GRCh38
rs369111058 Chr.19: 1990745-1990745 on Build GRCh38
rs1019264018 Chr.19: 1990639-1990639 on Build GRCh38

View Full Product Details