Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 98772977-98773246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKS1B
Entrez Gene ID
56899
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Gene Aliases
AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2
UniGene
Hs.506458
Gene Chromosome Location
Chr.12: 98729904-99984727 on Build GRCh38
Transcripts
NM_001204065.1 NM_001204066.1 NM_001204067.1 NM_001204068.1 NM_001204069.1 NM_001204070.1 NM_001204079.1 NM_001204080.1 NM_001204081.1 NM_020140.3 NM_152788.4 NM_181670.3 XM_005269028.4 XM_005269029.4 XM_005269032.3 XM_006719504.3 XM_006719505.3 XM_006719506.3 XM_006719507.3 XM_006719508.3 XM_006719509.3 XM_006719510.3 XM_006719512.3 XM_006719513.3 XM_006719514.3 XM_011538571.2 XM_017019651.1 XM_017019652.1 XM_017019653.1 XM_017019654.1 XM_017019655.1 XM_017019656.1 XM_017019657.1 XM_017019658.1 XM_017019659.1 XM_017019660.1 XM_017019661.1 XM_017019662.1 XM_017019663.1 XM_017019664.1 XM_017019665.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4489357 c.1219C>T p.H407Y Chr.12: 98773077-98773077 on Build GRCh38
5421664 c.962G>A p.R321H Chr.12: 98773154-98773154 on Build GRCh38
7299420 c.3409G>T p.A1137S Chr.12: 98773137-98773137 on Build GRCh38
3772761 c.1053T>C p.H351H Chr.12: 98773063-98773063 on Build GRCh38
6880251 c.1050T>A p.C350* Chr.12: 98773066-98773066 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752909343 Chr.12: 98773032-98773032 on Build GRCh38
rs374679636 Chr.12: 98773218-98773218 on Build GRCh38
rs752964424 Chr.12: 98773186-98773186 on Build GRCh38
rs759880783 Chr.12: 98773176-98773176 on Build GRCh38
rs751878396 Chr.12: 98773009-98773009 on Build GRCh38
rs755307256 Chr.12: 98773010-98773010 on Build GRCh38
rs774905244 Chr.12: 98773164-98773164 on Build GRCh38
rs769524758 Chr.12: 98773147-98773147 on Build GRCh38
rs758298431 Chr.12: 98773067-98773067 on Build GRCh38
rs747782639 Chr.12: 98773137-98773137 on Build GRCh38
rs763579194 Chr.12: 98773003-98773003 on Build GRCh38
rs762462536 Chr.12: 98773151-98773151 on Build GRCh38
rs1021593806 Chr.12: 98773096-98773096 on Build GRCh38
rs756349950 Chr.12: 98773205-98773205 on Build GRCh38
rs766872246 Chr.12: 98773007-98773007 on Build GRCh38
rs766813244 Chr.12: 98773152-98773152 on Build GRCh38
rs753919141 Chr.12: 98773219-98773219 on Build GRCh38
rs749316124 Chr.12: 98773053-98773053 on Build GRCh38
rs767890319 Chr.12: 98773178-98773178 on Build GRCh38
rs776464241 Chr.12: 98773112-98773112 on Build GRCh38
rs965673606 Chr.12: 98773155-98773155 on Build GRCh38
rs377466216 Chr.12: 98773072-98773072 on Build GRCh38
rs753366903 Chr.12: 98773032-98773032 on Build GRCh38
rs530191569 Chr.12: 98773080-98773080 on Build GRCh38
rs772883800 Chr.12: 98773150-98773150 on Build GRCh38
rs770001743 Chr.12: 98773177-98773177 on Build GRCh38
rs2287574 Chr.12: 98773027-98773027 on Build GRCh38
rs374589475 Chr.12: 98773045-98773045 on Build GRCh38
rs746291653 Chr.12: 98773065-98773065 on Build GRCh38
rs756223525 Chr.12: 98773034-98773034 on Build GRCh38
rs757201198 Chr.12: 98773205-98773206 on Build GRCh38
rs370342056 Chr.12: 98773105-98773105 on Build GRCh38

View Full Product Details