Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 9629707-9629976 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAS2R1
Entrez Gene ID
50834
Gene Name
taste 2 receptor member 1
Gene Aliases
T2R1, TRB7
UniGene
Hs.567492
Gene Chromosome Location
Chr.5: 9628997-9630351 on Build GRCh38
Transcripts
NM_019599.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4517208 c.298_299GG>AA p.G100N Chr.5: 9629734-9629735 on Build GRCh38
3142086 c.149G>T p.C50F Chr.5: 9629884-9629884 on Build GRCh38
1228518 c.221A>G p.E74G Chr.5: 9629812-9629812 on Build GRCh38
369339 c.240G>T p.A80A Chr.5: 9629793-9629793 on Build GRCh38
3142080 c.254T>C p.L85P Chr.5: 9629779-9629779 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760172148 Chr.5: 9629862-9629862 on Build GRCh38
rs201583253 Chr.5: 9629900-9629900 on Build GRCh38
rs768104638 Chr.5: 9629889-9629891 on Build GRCh38
rs368379193 Chr.5: 9629804-9629804 on Build GRCh38
rs748360046 Chr.5: 9629747-9629747 on Build GRCh38
rs760083467 Chr.5: 9629879-9629879 on Build GRCh38
rs200371021 Chr.5: 9629828-9629828 on Build GRCh38
rs753740244 Chr.5: 9629842-9629842 on Build GRCh38
rs757057177 Chr.5: 9629847-9629847 on Build GRCh38
rs775288698 Chr.5: 9629916-9629916 on Build GRCh38
rs138330930 Chr.5: 9629744-9629744 on Build GRCh38
rs750215367 Chr.5: 9629922-9629922 on Build GRCh38
rs139834398 Chr.5: 9629841-9629841 on Build GRCh38
rs139538133 Chr.5: 9629732-9629732 on Build GRCh38
rs571918935 Chr.5: 9629793-9629793 on Build GRCh38
rs752695852 Chr.5: 9629832-9629832 on Build GRCh38
rs375904658 Chr.5: 9629867-9629867 on Build GRCh38
rs1006092804 Chr.5: 9629945-9629945 on Build GRCh38
rs752888560 Chr.5: 9629894-9629894 on Build GRCh38
rs554466158 Chr.5: 9629878-9629878 on Build GRCh38
rs553576885 Chr.5: 9629727-9629727 on Build GRCh38
rs768417660 Chr.5: 9629860-9629860 on Build GRCh38
rs749005082 Chr.5: 9629759-9629759 on Build GRCh38
rs367652541 Chr.5: 9629895-9629895 on Build GRCh38
rs749941550 Chr.5: 9629845-9629845 on Build GRCh38
rs775030634 Chr.5: 9629779-9629779 on Build GRCh38
rs763291783 Chr.5: 9629807-9629807 on Build GRCh38
rs758432177 Chr.5: 9629930-9629930 on Build GRCh38
rs748340191 Chr.5: 9629797-9629797 on Build GRCh38
rs767878843 Chr.5: 9629882-9629882 on Build GRCh38
rs140220629 Chr.5: 9629872-9629872 on Build GRCh38
rs375037872 Chr.5: 9629800-9629800 on Build GRCh38
rs763312142 Chr.5: 9629875-9629875 on Build GRCh38
rs755882569 Chr.5: 9629751-9629751 on Build GRCh38
rs746500904 Chr.5: 9629857-9629857 on Build GRCh38
rs145857461 Chr.5: 9629854-9629854 on Build GRCh38
rs1046182747 Chr.5: 9629946-9629946 on Build GRCh38
rs372703858 Chr.5: 9629835-9629835 on Build GRCh38
rs370954622 Chr.5: 9629794-9629794 on Build GRCh38
rs777671671 Chr.5: 9629758-9629758 on Build GRCh38
rs753142882 Chr.5: 9629911-9629912 on Build GRCh38
rs2234231 Chr.5: 9629905-9629905 on Build GRCh38
rs868678343 Chr.5: 9629890-9629890 on Build GRCh38
rs745381132 Chr.5: 9629770-9629770 on Build GRCh38
rs749312242 Chr.5: 9629861-9629861 on Build GRCh38
rs149835603 Chr.5: 9629853-9629853 on Build GRCh38
rs769964745 Chr.5: 9629821-9629821 on Build GRCh38
rs771007431 Chr.5: 9629904-9629904 on Build GRCh38
rs73743124 Chr.5: 9629736-9629736 on Build GRCh38
rs150643783 Chr.5: 9629810-9629810 on Build GRCh38
rs771638808 Chr.5: 9629773-9629773 on Build GRCh38
rs757147182 Chr.5: 9629912-9629912 on Build GRCh38
rs745758419 Chr.5: 9629849-9629849 on Build GRCh38
rs987829477 Chr.5: 9629787-9629787 on Build GRCh38
rs200282241 Chr.5: 9629767-9629767 on Build GRCh38
rs901174542 Chr.5: 9629897-9629897 on Build GRCh38
rs767583293 Chr.5: 9629830-9629830 on Build GRCh38
rs866156814 Chr.5: 9629836-9629836 on Build GRCh38
rs751997597 Chr.5: 9629814-9629814 on Build GRCh38
rs755283447 Chr.5: 9629733-9629733 on Build GRCh38
rs771054326 Chr.5: 9629763-9629763 on Build GRCh38
rs778995507 Chr.5: 9629848-9629848 on Build GRCh38

View Full Product Details