Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 18941538-18941807 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAXO1
Entrez Gene ID
158297
Gene Name
stabilizer of axonemal microtubules 1
Gene Aliases
C9orf138, FAM154A
UniGene
Hs.731593
Gene Chromosome Location
Chr.9: 18927660-19049354 on Build GRCh38
Transcripts
NM_001287049.1 NM_001287050.1 NM_153707.3 XM_011517744.1 XM_011517745.1 XM_011517746.2 XM_011517747.2 XM_017014314.1 XM_017014315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3656445 c.305T>A p.L102H Chr.9: 18941753-18941753 on Build GRCh38
3656444 c.333C>T p.P111P Chr.9: 18941725-18941725 on Build GRCh38
608555 c.391G>A p.D131N Chr.9: 18941667-18941667 on Build GRCh38
6473073 c.374G>T p.S125I Chr.9: 18941684-18941684 on Build GRCh38
234693 c.276C>T p.F92F Chr.9: 18941782-18941782 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199758996 Chr.9: 18941742-18941742 on Build GRCh38
rs960701315 Chr.9: 18941722-18941722 on Build GRCh38
rs780451760 Chr.9: 18941706-18941706 on Build GRCh38
rs61743636 Chr.9: 18941747-18941747 on Build GRCh38
rs750763382 Chr.9: 18941611-18941611 on Build GRCh38
rs61743637 Chr.9: 18941749-18941749 on Build GRCh38
rs778039622 Chr.9: 18941739-18941739 on Build GRCh38
rs916381531 Chr.9: 18941584-18941584 on Build GRCh38
rs781711386 Chr.9: 18941608-18941608 on Build GRCh38
rs200791962 Chr.9: 18941737-18941737 on Build GRCh38
rs745614839 Chr.9: 18941785-18941785 on Build GRCh38
rs754344498 Chr.9: 18941711-18941711 on Build GRCh38
rs190494721 Chr.9: 18941627-18941627 on Build GRCh38
rs34257010 Chr.9: 18941705-18941705 on Build GRCh38
rs765665750 Chr.9: 18941773-18941773 on Build GRCh38
rs201332442 Chr.9: 18941782-18941782 on Build GRCh38
rs369535708 Chr.9: 18941725-18941725 on Build GRCh38
rs34769002 Chr.9: 18941668-18941668 on Build GRCh38
rs749069610 Chr.9: 18941721-18941742 on Build GRCh38
rs757458262 Chr.9: 18941781-18941781 on Build GRCh38
rs923900168 Chr.9: 18941649-18941649 on Build GRCh38
rs778882191 Chr.9: 18941720-18941720 on Build GRCh38
rs756702121 Chr.9: 18941613-18941613 on Build GRCh38
rs772291032 Chr.9: 18941729-18941729 on Build GRCh38
rs772233893 Chr.9: 18941658-18941658 on Build GRCh38
rs769718845 Chr.9: 18941748-18941748 on Build GRCh38
rs142086584 Chr.9: 18941691-18941691 on Build GRCh38
rs780357419 Chr.9: 18941621-18941621 on Build GRCh38
rs35329037 Chr.9: 18941646-18941646 on Build GRCh38
rs750820696 Chr.9: 18941702-18941702 on Build GRCh38
rs752444050 Chr.9: 18941598-18941598 on Build GRCh38
rs1023741605 Chr.9: 18941744-18941744 on Build GRCh38
rs745895990 Chr.9: 18941703-18941703 on Build GRCh38
rs144484870 Chr.9: 18941667-18941667 on Build GRCh38
rs373134677 Chr.9: 18941580-18941580 on Build GRCh38
rs764792860 Chr.9: 18941595-18941595 on Build GRCh38
rs889923355 Chr.9: 18941690-18941690 on Build GRCh38
rs116651834 Chr.9: 18941750-18941750 on Build GRCh38
rs142202664 Chr.9: 18941767-18941767 on Build GRCh38
rs755455108 Chr.9: 18941772-18941772 on Build GRCh38
rs776493132 Chr.9: 18941674-18941674 on Build GRCh38
rs990652238 Chr.9: 18941576-18941576 on Build GRCh38
rs748401562 Chr.9: 18941656-18941656 on Build GRCh38
rs376556426 Chr.9: 18941677-18941677 on Build GRCh38
rs755473119 Chr.9: 18941712-18941712 on Build GRCh38
rs763781591 Chr.9: 18941698-18941698 on Build GRCh38
rs543682096 Chr.9: 18941583-18941583 on Build GRCh38
rs770759213 Chr.9: 18941673-18941673 on Build GRCh38
rs201860022 Chr.9: 18941660-18941660 on Build GRCh38
rs757351370 Chr.9: 18941637-18941637 on Build GRCh38
rs573501142 Chr.9: 18941734-18941734 on Build GRCh38
rs761502460 Chr.9: 18941760-18941760 on Build GRCh38
rs148609380 Chr.9: 18941688-18941688 on Build GRCh38
rs774203011 Chr.9: 18941754-18941754 on Build GRCh38
rs541329541 Chr.9: 18941776-18941776 on Build GRCh38
rs112030642 Chr.9: 18941746-18941746 on Build GRCh38
rs565492307 Chr.9: 18941603-18941603 on Build GRCh38
rs944255908 Chr.9: 18941662-18941662 on Build GRCh38
rs779021573 Chr.9: 18941630-18941630 on Build GRCh38
rs754956008 Chr.9: 18941721-18941721 on Build GRCh38
rs146125443 Chr.9: 18941777-18941777 on Build GRCh38
rs762713089 Chr.9: 18941697-18941697 on Build GRCh38

View Full Product Details