Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 97978811-97979084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MATN2
Entrez Gene ID
4147
Gene Name
matrilin 2
Gene Aliases
-
UniGene
Hs.189445
Gene Chromosome Location
Chr.8: 97869021-98036720 on Build GRCh38
Transcripts
NM_001317748.1 NM_002380.4 NM_030583.3 XM_005250920.1 XM_017013417.1 XM_017013418.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6394637 c.1066A>G p.K356E Chr.8: 97978993-97978993 on Build GRCh38
3652089 c.1023C>T p.S341S Chr.8: 97978950-97978950 on Build GRCh38
5979575 c.1065_1066insG p.K356fs*14 Chr.8: 97978992-97978993 on Build GRCh38
5875171 c.992G>A p.C331Y Chr.8: 97978919-97978919 on Build GRCh38
5738398 c.1066_1067insG p.K356fs*14 Chr.8: 97978993-97978994 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901986724 Chr.8: 97978859-97978859 on Build GRCh38
rs761238031 Chr.8: 97978886-97978886 on Build GRCh38
rs138153044 Chr.8: 97978940-97978940 on Build GRCh38
rs1869607 Chr.8: 97979053-97979053 on Build GRCh38
rs184482356 Chr.8: 97978930-97978930 on Build GRCh38
rs374514918 Chr.8: 97979027-97979027 on Build GRCh38
rs781076571 Chr.8: 97978960-97978960 on Build GRCh38
rs375963488 Chr.8: 97978962-97978962 on Build GRCh38
rs776639702 Chr.8: 97978851-97978851 on Build GRCh38
rs146982976 Chr.8: 97978901-97978901 on Build GRCh38
rs758617448 Chr.8: 97979024-97979024 on Build GRCh38
rs752724049 Chr.8: 97979043-97979043 on Build GRCh38
rs773924132 Chr.8: 97979007-97979010 on Build GRCh38
rs374391695 Chr.8: 97978915-97978915 on Build GRCh38
rs765855052 Chr.8: 97978872-97978872 on Build GRCh38
rs867578482 Chr.8: 97978978-97978978 on Build GRCh38
rs757417938 Chr.8: 97978893-97978893 on Build GRCh38
rs371119965 Chr.8: 97979007-97979007 on Build GRCh38
rs529618104 Chr.8: 97979035-97979035 on Build GRCh38
rs554256293 Chr.8: 97978889-97978889 on Build GRCh38
rs781311410 Chr.8: 97979042-97979042 on Build GRCh38
rs750647860 Chr.8: 97979019-97979019 on Build GRCh38
rs369817801 Chr.8: 97978914-97978914 on Build GRCh38
rs1869609 Chr.8: 97978993-97978993 on Build GRCh38
rs372424373 Chr.8: 97979000-97979000 on Build GRCh38
rs369159037 Chr.8: 97978862-97978862 on Build GRCh38
rs748949867 Chr.8: 97978970-97978970 on Build GRCh38
rs769500934 Chr.8: 97978855-97978855 on Build GRCh38
rs768560128 Chr.8: 97978847-97978847 on Build GRCh38
rs747737536 Chr.8: 97978852-97978852 on Build GRCh38
rs777495156 Chr.8: 97979058-97979058 on Build GRCh38
rs780058690 Chr.8: 97978929-97978929 on Build GRCh38
rs754212296 Chr.8: 97978891-97978891 on Build GRCh38
rs376481997 Chr.8: 97979001-97979001 on Build GRCh38
rs545594991 Chr.8: 97978965-97978965 on Build GRCh38
rs772550234 Chr.8: 97978857-97978857 on Build GRCh38
rs375244323 Chr.8: 97978839-97978839 on Build GRCh38
rs371269983 Chr.8: 97978883-97978883 on Build GRCh38
rs929476138 Chr.8: 97978865-97978865 on Build GRCh38
rs1869608 Chr.8: 97978995-97978995 on Build GRCh38
rs373486105 Chr.8: 97978982-97978982 on Build GRCh38
rs772890822 Chr.8: 97978968-97978968 on Build GRCh38

View Full Product Details