Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 733139-733364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM57A
Entrez Gene ID
79850
Gene Name
family with sequence similarity 57 member A
Gene Aliases
CT120
UniGene
Hs.154396
Gene Chromosome Location
Chr.17: 732546-742968 on Build GRCh38
Transcripts
NM_001318006.1 NM_001318007.1 NM_001318008.1 NM_024792.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4683090 c.166A>G p.I56V Chr.17: 733141-733141 on Build GRCh38
5981076 c.167T>C p.I56T Chr.17: 733142-733142 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs960433487 Chr.17: 733259-733259 on Build GRCh38
rs769651760 Chr.17: 733197-733197 on Build GRCh38
rs768550773 Chr.17: 733175-733175 on Build GRCh38
rs967961339 Chr.17: 733239-733239 on Build GRCh38
rs1034649948 Chr.17: 733342-733342 on Build GRCh38
rs761598561 Chr.17: 733191-733191 on Build GRCh38
rs567006141 Chr.17: 733252-733252 on Build GRCh38
rs999719803 Chr.17: 733338-733338 on Build GRCh38
rs960984033 Chr.17: 733190-733190 on Build GRCh38
rs7213108 Chr.17: 733210-733210 on Build GRCh38
rs767994589 Chr.17: 733229-733229 on Build GRCh38
rs148990429 Chr.17: 733304-733304 on Build GRCh38
rs941603022 Chr.17: 733335-733335 on Build GRCh38
rs907940818 Chr.17: 733256-733256 on Build GRCh38
rs759664137 Chr.17: 733215-733215 on Build GRCh38
rs763156553 Chr.17: 733202-733202 on Build GRCh38
rs2657623 Chr.17: 733290-733290 on Build GRCh38
rs889589074 Chr.17: 733318-733318 on Build GRCh38
rs927651306 Chr.17: 733159-733159 on Build GRCh38
rs769866306 Chr.17: 733192-733192 on Build GRCh38
rs982920657 Chr.17: 733327-733327 on Build GRCh38
rs980752620 Chr.17: 733249-733249 on Build GRCh38
rs570062439 Chr.17: 733326-733326 on Build GRCh38
rs910096124 Chr.17: 733330-733330 on Build GRCh38
rs540167069 Chr.17: 733301-733301 on Build GRCh38
rs565895079 Chr.17: 733300-733300 on Build GRCh38
rs892282141 Chr.17: 733195-733195 on Build GRCh38
rs1040367382 Chr.17: 733262-733262 on Build GRCh38
rs867704758 Chr.17: 733237-733237 on Build GRCh38
rs992299029 Chr.17: 733260-733260 on Build GRCh38
rs1022189533 Chr.17: 733231-733231 on Build GRCh38
rs901880807 Chr.17: 733286-733286 on Build GRCh38
rs368759793 Chr.17: 733205-733205 on Build GRCh38
rs773234177 Chr.17: 733201-733201 on Build GRCh38
rs776282878 Chr.17: 733189-733189 on Build GRCh38
rs549370437 Chr.17: 733255-733255 on Build GRCh38

View Full Product Details