Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 37522628-37522881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO10
Entrez Gene ID
26267
Gene Name
F-box protein 10
Gene Aliases
FBX10, PRMT11
UniGene
Hs.709527
Gene Chromosome Location
Chr.9: 37509920-37576459 on Build GRCh38
Transcripts
NM_012166.2 XM_005251439.4 XM_006716754.3 XM_017014618.1 XM_017014619.1

SNP Detail

SNP ID Chromosome Location
rs10973394 Chr.9: 37522675-37522675 on Build GRCh38
rs779913467 Chr.9: 37522837-37522837 on Build GRCh38
rs957003398 Chr.9: 37522726-37522726 on Build GRCh38
rs980669285 Chr.9: 37522861-37522861 on Build GRCh38
rs539580900 Chr.9: 37522817-37522817 on Build GRCh38
rs754995783 Chr.9: 37522862-37522862 on Build GRCh38
rs966919358 Chr.9: 37522655-37522655 on Build GRCh38
rs193296204 Chr.9: 37522668-37522668 on Build GRCh38
rs1036921528 Chr.9: 37522805-37522805 on Build GRCh38
rs758186106 Chr.9: 37522826-37522826 on Build GRCh38
rs953194962 Chr.9: 37522741-37522741 on Build GRCh38
rs989920846 Chr.9: 37522727-37522727 on Build GRCh38
rs961926629 Chr.9: 37522795-37522795 on Build GRCh38
rs971978651 Chr.9: 37522806-37522806 on Build GRCh38
rs377542171 Chr.9: 37522774-37522774 on Build GRCh38
rs12343402 Chr.9: 37522815-37522815 on Build GRCh38
rs765172172 Chr.9: 37522793-37522793 on Build GRCh38
rs939939138 Chr.9: 37522799-37522799 on Build GRCh38
rs374192200 Chr.9: 37522859-37522859 on Build GRCh38
rs984437030 Chr.9: 37522755-37522755 on Build GRCh38
rs921012396 Chr.9: 37522730-37522730 on Build GRCh38
rs761323274 Chr.9: 37522682-37522682 on Build GRCh38
rs374397106 Chr.9: 37522836-37522836 on Build GRCh38
rs756741985 Chr.9: 37522792-37522792 on Build GRCh38

View Full Product Details