Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 42770034-42770307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAAO
Entrez Gene ID
23498
Gene Name
3-hydroxyanthranilate 3,4-dioxygenase
Gene Aliases
3-HAO, HAO
UniGene
Hs.368805
Gene Chromosome Location
Chr.2: 42766198-42794359 on Build GRCh38
Transcripts
NM_012205.2 XM_005264230.3 XM_011532729.2 XM_011532730.2 XM_011532731.2 XM_017003717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
212871 c.459G>C p.Q153H Chr.2: 42770168-42770168 on Build GRCh38
442910 c.444C>T p.F148F Chr.2: 42770183-42770183 on Build GRCh38
4753817 c.476C>A p.P159H Chr.2: 42770151-42770151 on Build GRCh38
5620927 c.466A>T p.T156S Chr.2: 42770161-42770161 on Build GRCh38
442909 c.447C>T p.F149F Chr.2: 42770180-42770180 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374774681 Chr.2: 42770197-42770197 on Build GRCh38
rs372925453 Chr.2: 42770176-42770176 on Build GRCh38
rs754038457 Chr.2: 42770211-42770212 on Build GRCh38
rs751238695 Chr.2: 42770288-42770288 on Build GRCh38
rs527656756 Chr.2: 42770143-42770144 on Build GRCh38
rs1033095279 Chr.2: 42770204-42770204 on Build GRCh38
rs770973219 Chr.2: 42770219-42770219 on Build GRCh38
rs991789433 Chr.2: 42770068-42770068 on Build GRCh38
rs866117467 Chr.2: 42770164-42770164 on Build GRCh38
rs755657772 Chr.2: 42770169-42770169 on Build GRCh38
rs951718133 Chr.2: 42770245-42770245 on Build GRCh38
rs891859817 Chr.2: 42770214-42770214 on Build GRCh38
rs779772454 Chr.2: 42770183-42770183 on Build GRCh38
rs368790811 Chr.2: 42770244-42770244 on Build GRCh38
rs975006551 Chr.2: 42770058-42770058 on Build GRCh38
rs889147053 Chr.2: 42770227-42770227 on Build GRCh38
rs1019556866 Chr.2: 42770229-42770229 on Build GRCh38
rs757961797 Chr.2: 42770064-42770064 on Build GRCh38
rs534381889 Chr.2: 42770126-42770126 on Build GRCh38
rs757381641 Chr.2: 42770212-42770232 on Build GRCh38
rs776150246 Chr.2: 42770232-42770232 on Build GRCh38
rs758953283 Chr.2: 42770134-42770134 on Build GRCh38
rs147810491 Chr.2: 42770070-42770070 on Build GRCh38
rs1031761745 Chr.2: 42770284-42770284 on Build GRCh38
rs747123946 Chr.2: 42770215-42770215 on Build GRCh38
rs761259457 Chr.2: 42770180-42770180 on Build GRCh38
rs960248140 Chr.2: 42770067-42770067 on Build GRCh38
rs908937711 Chr.2: 42770144-42770144 on Build GRCh38
rs998987406 Chr.2: 42770256-42770256 on Build GRCh38
rs759932757 Chr.2: 42770123-42770123 on Build GRCh38
rs369185081 Chr.2: 42770066-42770066 on Build GRCh38
rs1056197847 Chr.2: 42770128-42770128 on Build GRCh38
rs754091154 Chr.2: 42770108-42770108 on Build GRCh38
rs768537500 Chr.2: 42770187-42770187 on Build GRCh38
rs565804501 Chr.2: 42770089-42770089 on Build GRCh38
rs776909901 Chr.2: 42770233-42770233 on Build GRCh38
rs749984965 Chr.2: 42770135-42770135 on Build GRCh38
rs764685666 Chr.2: 42770197-42770206 on Build GRCh38
rs141209245 Chr.2: 42770196-42770196 on Build GRCh38
rs184674290 Chr.2: 42770257-42770257 on Build GRCh38
rs753171294 Chr.2: 42770131-42770131 on Build GRCh38
rs1044985475 Chr.2: 42770155-42770155 on Build GRCh38
rs1012600882 Chr.2: 42770147-42770147 on Build GRCh38
rs772166124 Chr.2: 42770207-42770207 on Build GRCh38
rs748957756 Chr.2: 42770186-42770186 on Build GRCh38
rs907643153 Chr.2: 42770268-42770268 on Build GRCh38
rs192695179 Chr.2: 42770213-42770213 on Build GRCh38
rs969067136 Chr.2: 42770086-42770086 on Build GRCh38

View Full Product Details