Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 77165652-77165907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNG2
Entrez Gene ID
901
Gene Name
cyclin G2
Gene Aliases
-
UniGene
Hs.740456
Gene Chromosome Location
Chr.4: 77157204-77170060 on Build GRCh38
Transcripts
NM_004354.2 XM_011532398.1 XM_011532399.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1694582 c.962C>T p.S321F Chr.4: 77165851-77165851 on Build GRCh38
127570 c.948G>A p.E316E Chr.4: 77165837-77165837 on Build GRCh38
7096054 c.996C>T p.F332F Chr.4: 77165885-77165885 on Build GRCh38
1431127 c.938G>A p.C313Y Chr.4: 77165827-77165827 on Build GRCh38
80986 c.1002C>T p.F334F Chr.4: 77165891-77165891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201736578 Chr.4: 77165821-77165821 on Build GRCh38
rs1037012415 Chr.4: 77165862-77165862 on Build GRCh38
rs768781122 Chr.4: 77165860-77165860 on Build GRCh38
rs758354196 Chr.4: 77165851-77165851 on Build GRCh38
rs769721809 Chr.4: 77165796-77165796 on Build GRCh38
rs4150092 Chr.4: 77165747-77165747 on Build GRCh38
rs925487820 Chr.4: 77165738-77165738 on Build GRCh38
rs774852045 Chr.4: 77165807-77165807 on Build GRCh38
rs754544955 Chr.4: 77165842-77165842 on Build GRCh38
rs918080743 Chr.4: 77165684-77165684 on Build GRCh38
rs763049090 Chr.4: 77165798-77165798 on Build GRCh38
rs777614298 Chr.4: 77165771-77165771 on Build GRCh38
rs780254344 Chr.4: 77165859-77165859 on Build GRCh38
rs771780763 Chr.4: 77165769-77165769 on Build GRCh38
rs564789326 Chr.4: 77165687-77165687 on Build GRCh38
rs759167996 Chr.4: 77165866-77165867 on Build GRCh38
rs753502032 Chr.4: 77165839-77165839 on Build GRCh38
rs933190481 Chr.4: 77165871-77165871 on Build GRCh38
rs1049732503 Chr.4: 77165808-77165808 on Build GRCh38
rs761745402 Chr.4: 77165775-77165776 on Build GRCh38
rs528949702 Chr.4: 77165789-77165789 on Build GRCh38
rs766152961 Chr.4: 77165831-77165831 on Build GRCh38
rs754489986 Chr.4: 77165751-77165751 on Build GRCh38
rs369436663 Chr.4: 77165853-77165853 on Build GRCh38
rs374181852 Chr.4: 77165864-77165864 on Build GRCh38
rs768652357 Chr.4: 77165805-77165805 on Build GRCh38
rs563766657 Chr.4: 77165876-77165876 on Build GRCh38
rs557271897 Chr.4: 77165689-77165689 on Build GRCh38
rs752638436 Chr.4: 77165849-77165849 on Build GRCh38
rs769979003 Chr.4: 77165775-77165775 on Build GRCh38
rs957845609 Chr.4: 77165715-77165715 on Build GRCh38
rs748055630 Chr.4: 77165768-77165768 on Build GRCh38
rs189401726 Chr.4: 77165856-77165856 on Build GRCh38
rs762432430 Chr.4: 77165807-77165808 on Build GRCh38
rs762368414 Chr.4: 77165810-77165810 on Build GRCh38
rs774483140 Chr.4: 77165867-77165867 on Build GRCh38
rs767835755 Chr.4: 77165812-77165812 on Build GRCh38
rs541522203 Chr.4: 77165846-77165846 on Build GRCh38
rs75208252 Chr.4: 77165873-77165873 on Build GRCh38
rs909946356 Chr.4: 77165843-77165843 on Build GRCh38
rs779026050 Chr.4: 77165761-77165761 on Build GRCh38
rs201197612 Chr.4: 77165854-77165854 on Build GRCh38
rs4150093 Chr.4: 77165772-77165772 on Build GRCh38
rs772770660 Chr.4: 77165799-77165800 on Build GRCh38
rs776342196 Chr.4: 77165719-77165719 on Build GRCh38
rs982340746 Chr.4: 77165820-77165820 on Build GRCh38
rs978366503 Chr.4: 77165732-77165732 on Build GRCh38
rs751190368 Chr.4: 77165829-77165833 on Build GRCh38
rs765868965 Chr.4: 77165810-77165811 on Build GRCh38
rs761556512 Chr.4: 77165728-77165728 on Build GRCh38

View Full Product Details