Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155253530-155253803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM189B
Entrez Gene ID
10712
Gene Name
family with sequence similarity 189 member B
Gene Aliases
C1orf2, COTE1
UniGene
Hs.348308
Gene Chromosome Location
Chr.1: 155247205-155255483 on Build GRCh38
Transcripts
NM_001267608.1 NM_006589.2 NM_198264.1 XM_005244845.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
388870 c.620C>T p.T207M Chr.1: 155253626-155253626 on Build GRCh38
6659159 c.538C>T p.L180F Chr.1: 155253708-155253708 on Build GRCh38
3802097 c.541A>C p.T181P Chr.1: 155253705-155253705 on Build GRCh38
896836 c.514-1G>T p.? Chr.1: 155253733-155253733 on Build GRCh38
3740690 c.552C>T p.A184A Chr.1: 155253694-155253694 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991456752 Chr.1: 155253566-155253566 on Build GRCh38
rs149844175 Chr.1: 155253693-155253693 on Build GRCh38
rs368091936 Chr.1: 155253597-155253597 on Build GRCh38
rs948431091 Chr.1: 155253672-155253672 on Build GRCh38
rs745312643 Chr.1: 155253554-155253554 on Build GRCh38
rs774206973 Chr.1: 155253717-155253717 on Build GRCh38
rs147222012 Chr.1: 155253598-155253598 on Build GRCh38
rs756372703 Chr.1: 155253626-155253626 on Build GRCh38
rs112532910 Chr.1: 155253710-155253710 on Build GRCh38
rs765034073 Chr.1: 155253610-155253610 on Build GRCh38
rs780923272 Chr.1: 155253751-155253751 on Build GRCh38
rs768292729 Chr.1: 155253743-155253743 on Build GRCh38
rs750385680 Chr.1: 155253622-155253622 on Build GRCh38
rs781471922 Chr.1: 155253634-155253634 on Build GRCh38
rs779113167 Chr.1: 155253598-155253598 on Build GRCh38
rs757788503 Chr.1: 155253582-155253582 on Build GRCh38
rs780737064 Chr.1: 155253720-155253720 on Build GRCh38
rs139976204 Chr.1: 155253662-155253662 on Build GRCh38
rs867685856 Chr.1: 155253695-155253695 on Build GRCh38
rs375507400 Chr.1: 155253638-155253638 on Build GRCh38
rs958783679 Chr.1: 155253557-155253557 on Build GRCh38
rs760462627 Chr.1: 155253593-155253593 on Build GRCh38
rs779759611 Chr.1: 155253644-155253644 on Build GRCh38
rs77452382 Chr.1: 155253718-155253718 on Build GRCh38
rs141407917 Chr.1: 155253657-155253657 on Build GRCh38
rs755637710 Chr.1: 155253748-155253752 on Build GRCh38
rs142048704 Chr.1: 155253707-155253707 on Build GRCh38
rs774897854 Chr.1: 155253676-155253676 on Build GRCh38
rs563495253 Chr.1: 155253576-155253606 on Build GRCh38
rs768382581 Chr.1: 155253648-155253648 on Build GRCh38
rs967203434 Chr.1: 155253604-155253604 on Build GRCh38
rs773104190 Chr.1: 155253780-155253780 on Build GRCh38
rs774948480 Chr.1: 155253552-155253552 on Build GRCh38
rs746630229 Chr.1: 155253646-155253646 on Build GRCh38
rs932131917 Chr.1: 155253639-155253639 on Build GRCh38
rs780964889 Chr.1: 155253631-155253632 on Build GRCh38
rs368433683 Chr.1: 155253741-155253741 on Build GRCh38
rs776607270 Chr.1: 155253599-155253599 on Build GRCh38
rs771619516 Chr.1: 155253570-155253570 on Build GRCh38
rs779392829 Chr.1: 155253708-155253708 on Build GRCh38
rs367691667 Chr.1: 155253659-155253659 on Build GRCh38
rs746455206 Chr.1: 155253636-155253636 on Build GRCh38
rs371935188 Chr.1: 155253666-155253666 on Build GRCh38
rs564062941 Chr.1: 155253574-155253574 on Build GRCh38
rs747759668 Chr.1: 155253645-155253647 on Build GRCh38
rs750845959 Chr.1: 155253598-155253599 on Build GRCh38
rs769460735 Chr.1: 155253768-155253768 on Build GRCh38
rs754944228 Chr.1: 155253625-155253625 on Build GRCh38
rs754571474 Chr.1: 155253606-155253607 on Build GRCh38
rs142840486 Chr.1: 155253767-155253767 on Build GRCh38
rs754157328 Chr.1: 155253633-155253633 on Build GRCh38
rs754290276 Chr.1: 155253576-155253576 on Build GRCh38
rs139864823 Chr.1: 155253649-155253649 on Build GRCh38
rs754065628 Chr.1: 155253700-155253700 on Build GRCh38
rs764247381 Chr.1: 155253694-155253694 on Build GRCh38
rs147288480 Chr.1: 155253678-155253678 on Build GRCh38

View Full Product Details