Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 51280874-51281099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM124A
Entrez Gene ID
220108
Gene Name
family with sequence similarity 124 member A
Gene Aliases
-
UniGene
Hs.71913
Gene Chromosome Location
Chr.13: 51222328-51284241 on Build GRCh38
Transcripts
NM_001242312.1 NM_145019.3 XM_011534978.2 XM_017020419.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4911052 c.1410C>T p.P470P Chr.13: 51280917-51280917 on Build GRCh38
6139071 c.1477C>T p.H493Y Chr.13: 51280984-51280984 on Build GRCh38
4527870 c.1496G>A p.G499E Chr.13: 51281003-51281003 on Build GRCh38
1300244 c.1592G>T p.R531L Chr.13: 51281099-51281099 on Build GRCh38
947938 c.1419C>A p.F473L Chr.13: 51280926-51280926 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764691984 Chr.13: 51280946-51280946 on Build GRCh38
rs776764205 Chr.13: 51281034-51281034 on Build GRCh38
rs750523295 Chr.13: 51280965-51280965 on Build GRCh38
rs773890927 Chr.13: 51280924-51280924 on Build GRCh38
rs770502031 Chr.13: 51281014-51281014 on Build GRCh38
rs755357233 Chr.13: 51280972-51280972 on Build GRCh38
rs369888482 Chr.13: 51280934-51280934 on Build GRCh38
rs749738902 Chr.13: 51280968-51280968 on Build GRCh38
rs748080141 Chr.13: 51280955-51280955 on Build GRCh38
rs927790247 Chr.13: 51281027-51281027 on Build GRCh38
rs74087446 Chr.13: 51280978-51280978 on Build GRCh38
rs764976880 Chr.13: 51281039-51281039 on Build GRCh38
rs767810486 Chr.13: 51281073-51281073 on Build GRCh38
rs772701284 Chr.13: 51280955-51280955 on Build GRCh38
rs772695251 Chr.13: 51280992-51280992 on Build GRCh38
rs199695346 Chr.13: 51281059-51281059 on Build GRCh38
rs771549646 Chr.13: 51280935-51280935 on Build GRCh38
rs912534192 Chr.13: 51281008-51281008 on Build GRCh38
rs756201060 Chr.13: 51281081-51281081 on Build GRCh38
rs978087114 Chr.13: 51280918-51280918 on Build GRCh38
rs939164230 Chr.13: 51281058-51281058 on Build GRCh38
rs201425404 Chr.13: 51280967-51280967 on Build GRCh38
rs2297969 Chr.13: 51280956-51280956 on Build GRCh38
rs775329106 Chr.13: 51281040-51281040 on Build GRCh38
rs73492203 Chr.13: 51280907-51280907 on Build GRCh38
rs771738151 Chr.13: 51280984-51280984 on Build GRCh38
rs185038718 Chr.13: 51280950-51280950 on Build GRCh38
rs767542969 Chr.13: 51280961-51280961 on Build GRCh38
rs749683400 Chr.13: 51280903-51280903 on Build GRCh38
rs555897503 Chr.13: 51280951-51280951 on Build GRCh38
rs772751758 Chr.13: 51280943-51280943 on Build GRCh38
rs887008248 Chr.13: 51281062-51281062 on Build GRCh38
rs202111589 Chr.13: 51280945-51280945 on Build GRCh38
rs200189512 Chr.13: 51281035-51281035 on Build GRCh38
rs757466454 Chr.13: 51280957-51280957 on Build GRCh38
rs746552464 Chr.13: 51280996-51280996 on Build GRCh38

View Full Product Details