Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 12704513-12704738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPPED1
Entrez Gene ID
55313
Gene Name
calcineurin like phosphoesterase domain containing 1
Gene Aliases
CSTP1
UniGene
Hs.460002
Gene Chromosome Location
Chr.16: 12659799-12803887 on Build GRCh38
Transcripts
NM_001099455.1 NM_018340.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1640394 c.683G>A p.R228Q Chr.16: 12704656-12704656 on Build GRCh38
460609 c.640G>A p.D214N Chr.16: 12704699-12704699 on Build GRCh38
5952151 c.625T>C p.F209L Chr.16: 12704714-12704714 on Build GRCh38
7266698 c.648C>T p.D216D Chr.16: 12704691-12704691 on Build GRCh38
5957142 c.649G>A p.D217N Chr.16: 12704690-12704690 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747916970 Chr.16: 12704609-12704609 on Build GRCh38
rs937703008 Chr.16: 12704542-12704542 on Build GRCh38
rs200745290 Chr.16: 12704697-12704697 on Build GRCh38
rs890583445 Chr.16: 12704580-12704580 on Build GRCh38
rs375350802 Chr.16: 12704605-12704605 on Build GRCh38
rs749775229 Chr.16: 12704589-12704589 on Build GRCh38
rs758424847 Chr.16: 12704659-12704659 on Build GRCh38
rs758514558 Chr.16: 12704718-12704718 on Build GRCh38
rs373177938 Chr.16: 12704579-12704579 on Build GRCh38
rs140031042 Chr.16: 12704582-12704582 on Build GRCh38
rs1027629262 Chr.16: 12704644-12704644 on Build GRCh38
rs569794425 Chr.16: 12704554-12704554 on Build GRCh38
rs780190524 Chr.16: 12704660-12704660 on Build GRCh38
rs754364791 Chr.16: 12704633-12704633 on Build GRCh38
rs757778243 Chr.16: 12704645-12704645 on Build GRCh38
rs764518637 Chr.16: 12704628-12704628 on Build GRCh38
rs780178007 Chr.16: 12704719-12704719 on Build GRCh38
rs752369308 Chr.16: 12704626-12704626 on Build GRCh38
rs374136858 Chr.16: 12704627-12704627 on Build GRCh38
rs774004567 Chr.16: 12704619-12704619 on Build GRCh38
rs753037267 Chr.16: 12704578-12704578 on Build GRCh38
rs200125670 Chr.16: 12704624-12704624 on Build GRCh38
rs201083958 Chr.16: 12704612-12704612 on Build GRCh38
rs774379222 Chr.16: 12704690-12704690 on Build GRCh38
rs550379364 Chr.16: 12704551-12704551 on Build GRCh38
rs926520262 Chr.16: 12704540-12704540 on Build GRCh38
rs765757532 Chr.16: 12704704-12704704 on Build GRCh38
rs771021460 Chr.16: 12704687-12704687 on Build GRCh38
rs762127968 Chr.16: 12704700-12704700 on Build GRCh38
rs747224066 Chr.16: 12704668-12704668 on Build GRCh38
rs757391292 Chr.16: 12704590-12704590 on Build GRCh38
rs199536895 Chr.16: 12704648-12704648 on Build GRCh38
rs759075092 Chr.16: 12704623-12704623 on Build GRCh38
rs773071674 Chr.16: 12704683-12704683 on Build GRCh38
rs534616041 Chr.16: 12704684-12704684 on Build GRCh38
rs754208671 Chr.16: 12704698-12704698 on Build GRCh38
rs763749898 Chr.16: 12704635-12704635 on Build GRCh38
rs759163128 Chr.16: 12704691-12704691 on Build GRCh38
rs767019120 Chr.16: 12704692-12704692 on Build GRCh38
rs760275214 Chr.16: 12704577-12704577 on Build GRCh38
rs565629915 Chr.16: 12704674-12704674 on Build GRCh38
rs775067554 Chr.16: 12704696-12704696 on Build GRCh38
rs931139805 Chr.16: 12704587-12704587 on Build GRCh38
rs377450216 Chr.16: 12704606-12704606 on Build GRCh38
rs778947063 Chr.16: 12704598-12704598 on Build GRCh38
rs959295487 Chr.16: 12704699-12704699 on Build GRCh38
rs767214778 Chr.16: 12704574-12704574 on Build GRCh38
rs773062958 Chr.16: 12704613-12704613 on Build GRCh38
rs750908927 Chr.16: 12704712-12704712 on Build GRCh38
rs753355629 Chr.16: 12704656-12704656 on Build GRCh38
rs766362537 Chr.16: 12704614-12704614 on Build GRCh38
rs914993415 Chr.16: 12704552-12704552 on Build GRCh38
rs775739910 Chr.16: 12704608-12704608 on Build GRCh38

View Full Product Details