Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 722561-722830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC78
Entrez Gene ID
124093
Gene Name
coiled-coil domain containing 78
Gene Aliases
C16orf25, CNM4, JFP10, hsCCDC78
UniGene
Hs.381943
Gene Chromosome Location
Chr.16: 722578-726880 on Build GRCh38
Transcripts
NM_001031737.2 XM_006720838.1 XM_006720843.3 XM_011522356.1 XM_011522357.1 XM_011522358.2 XM_011522359.1 XM_011522360.1 XM_011522361.1 XM_011522362.1 XM_011522363.1 XM_011522364.1 XM_011522365.1 XM_011522366.1 XM_011522367.1 XM_011522368.1 XM_011522369.1 XM_011522370.1 XM_011522371.2 XM_017022929.1 XM_017022930.1 XM_017022931.1 XM_017022932.1 NM_001271285.1 NM_023933.2

Gene Symbol
FAM173A
Entrez Gene ID
65990
Gene Name
family with sequence similarity 173 member A
Gene Aliases
C16orf24
UniGene
Hs.166244
Gene Chromosome Location
Chr.16: 720581-722590 on Build GRCh38
Transcripts
NM_001031737.2 XM_006720838.1 XM_006720843.3 XM_011522356.1 XM_011522357.1 XM_011522358.2 XM_011522359.1 XM_011522360.1 XM_011522361.1 XM_011522362.1 XM_011522363.1 XM_011522364.1 XM_011522365.1 XM_011522366.1 XM_011522367.1 XM_011522368.1 XM_011522369.1 XM_011522370.1 XM_011522371.2 XM_017022929.1 XM_017022930.1 XM_017022931.1 XM_017022932.1 NM_001271285.1 NM_023933.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4998526 c.1304G>T p.S435I Chr.16: 722783-722783 on Build GRCh38
6622449 c.1308G>A p.T436T Chr.16: 722779-722779 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748114954 Chr.16: 722784-722784 on Build GRCh38
rs766665690 Chr.16: 722691-722691 on Build GRCh38
rs775532654 Chr.16: 722602-722602 on Build GRCh38
rs779445722 Chr.16: 722638-722638 on Build GRCh38
rs753986011 Chr.16: 722693-722693 on Build GRCh38
rs553588726 Chr.16: 722645-722646 on Build GRCh38
rs748762375 Chr.16: 722759-722759 on Build GRCh38
rs779726829 Chr.16: 722756-722756 on Build GRCh38
rs374237828 Chr.16: 722808-722808 on Build GRCh38
rs751984462 Chr.16: 722792-722792 on Build GRCh38
rs753983692 Chr.16: 722631-722631 on Build GRCh38
rs757695991 Chr.16: 722795-722795 on Build GRCh38
rs770638452 Chr.16: 722654-722654 on Build GRCh38
rs138151643 Chr.16: 722783-722783 on Build GRCh38
rs764699064 Chr.16: 722790-722790 on Build GRCh38
rs146983005 Chr.16: 722768-722768 on Build GRCh38
rs745421264 Chr.16: 722674-722674 on Build GRCh38
rs769508152 Chr.16: 722750-722750 on Build GRCh38
rs541439427 Chr.16: 722689-722689 on Build GRCh38
rs762887103 Chr.16: 722606-722606 on Build GRCh38
rs775569629 Chr.16: 722780-722780 on Build GRCh38
rs1024185182 Chr.16: 722623-722623 on Build GRCh38
rs777451483 Chr.16: 722646-722650 on Build GRCh38
rs113597894 Chr.16: 722644-722644 on Build GRCh38
rs780665863 Chr.16: 722664-722664 on Build GRCh38
rs765297346 Chr.16: 722724-722724 on Build GRCh38
rs768749790 Chr.16: 722761-722761 on Build GRCh38
rs771752261 Chr.16: 722766-722766 on Build GRCh38
rs1048490947 Chr.16: 722657-722657 on Build GRCh38
rs758827941 Chr.16: 722646-722646 on Build GRCh38
rs770423446 Chr.16: 722593-722593 on Build GRCh38
rs757527717 Chr.16: 722735-722735 on Build GRCh38
rs906056013 Chr.16: 722730-722730 on Build GRCh38
rs574166000 Chr.16: 722649-722649 on Build GRCh38
rs765383192 Chr.16: 722779-722779 on Build GRCh38
rs977147162 Chr.16: 722642-722642 on Build GRCh38
rs776216360 Chr.16: 722599-722599 on Build GRCh38
rs772403335 Chr.16: 722588-722588 on Build GRCh38
rs775622579 Chr.16: 722680-722680 on Build GRCh38
rs761477432 Chr.16: 722690-722690 on Build GRCh38
rs562902898 Chr.16: 722700-722700 on Build GRCh38
rs760967870 Chr.16: 722609-722609 on Build GRCh38
rs746669448 Chr.16: 722592-722592 on Build GRCh38
rs185369512 Chr.16: 722762-722762 on Build GRCh38
rs769881632 Chr.16: 722601-722601 on Build GRCh38
rs755021500 Chr.16: 722637-722637 on Build GRCh38
rs759758193 Chr.16: 722775-722775 on Build GRCh38
rs201006875 Chr.16: 722797-722797 on Build GRCh38
rs201548248 Chr.16: 722788-722788 on Build GRCh38
rs971716715 Chr.16: 722626-722626 on Build GRCh38
rs745499090 Chr.16: 722748-722748 on Build GRCh38
rs780944551 Chr.16: 722740-722740 on Build GRCh38
rs532033150 Chr.16: 722725-722725 on Build GRCh38
rs940064441 Chr.16: 722675-722675 on Build GRCh38
rs747277625 Chr.16: 722652-722652 on Build GRCh38
rs768661834 Chr.16: 722688-722688 on Build GRCh38
rs751983703 Chr.16: 722733-722733 on Build GRCh38
rs753189763 Chr.16: 722640-722640 on Build GRCh38
rs769402064 Chr.16: 722678-722678 on Build GRCh38
rs541205240 Chr.16: 722608-722608 on Build GRCh38
rs772908784 Chr.16: 722590-722590 on Build GRCh38
rs180696429 Chr.16: 722600-722600 on Build GRCh38
rs763980269 Chr.16: 722607-722607 on Build GRCh38
rs748024891 Chr.16: 722764-722764 on Build GRCh38
rs559050074 Chr.16: 722627-722627 on Build GRCh38
rs778093019 Chr.16: 722727-722727 on Build GRCh38

View Full Product Details