Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 110051311-110051572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL6
Entrez Gene ID
79071
Gene Name
ELOVL fatty acid elongase 6
Gene Aliases
FACE, FAE, LCE
UniGene
Hs.412939
Gene Chromosome Location
Chr.4: 110049073-110199199 on Build GRCh38
Transcripts
NM_001130721.1 NM_024090.2 XM_011532233.2 XM_011532234.2 XM_011532235.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3006413 c.614A>G p.Q205R Chr.4: 110051522-110051522 on Build GRCh38
4121356 c.656G>A p.W219* Chr.4: 110051480-110051480 on Build GRCh38
1671205 c.727C>A p.L243I Chr.4: 110051409-110051409 on Build GRCh38
3775476 c.774G>A p.M258I Chr.4: 110051362-110051362 on Build GRCh38
1327987 c.655T>C p.W219R Chr.4: 110051481-110051481 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747887883 Chr.4: 110051425-110051425 on Build GRCh38
rs781014373 Chr.4: 110051409-110051409 on Build GRCh38
rs776031370 Chr.4: 110051491-110051491 on Build GRCh38
rs374061960 Chr.4: 110051370-110051370 on Build GRCh38
rs774835695 Chr.4: 110051345-110051345 on Build GRCh38
rs142637943 Chr.4: 110051371-110051371 on Build GRCh38
rs749170689 Chr.4: 110051441-110051441 on Build GRCh38
rs140803213 Chr.4: 110051482-110051482 on Build GRCh38
rs768764970 Chr.4: 110051341-110051341 on Build GRCh38
rs757865491 Chr.4: 110051363-110051363 on Build GRCh38
rs764708158 Chr.4: 110051502-110051502 on Build GRCh38
rs749955675 Chr.4: 110051360-110051360 on Build GRCh38
rs771110566 Chr.4: 110051446-110051446 on Build GRCh38
rs148477275 Chr.4: 110051350-110051350 on Build GRCh38
rs375827628 Chr.4: 110051427-110051427 on Build GRCh38
rs754449355 Chr.4: 110051503-110051503 on Build GRCh38
rs764659085 Chr.4: 110051351-110051351 on Build GRCh38
rs759855141 Chr.4: 110051481-110051481 on Build GRCh38
rs761051529 Chr.4: 110051497-110051497 on Build GRCh38
rs774265198 Chr.4: 110051464-110051464 on Build GRCh38
rs150982821 Chr.4: 110051373-110051373 on Build GRCh38
rs370638889 Chr.4: 110051342-110051342 on Build GRCh38
rs373791923 Chr.4: 110051506-110051506 on Build GRCh38

View Full Product Details