Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160862514-160862775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD244
Entrez Gene ID
51744
Gene Name
CD244 molecule
Gene Aliases
2B4, NAIL, NKR2B4, Nmrk, SLAMF4
UniGene
Hs.157872
Gene Chromosome Location
Chr.1: 160830160-160862902 on Build GRCh38
Transcripts
NM_001166663.1 NM_001166664.1 NM_016382.3 XM_011509621.2 XM_011509622.2 XM_011509623.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4518777 c.7_8GG>AA p.G3K Chr.1: 160862670-160862671 on Build GRCh38
4518779 c.7_8GG>AA p.G3K Chr.1: 160862670-160862671 on Build GRCh38
6340721 c.35T>A p.L12Q Chr.1: 160862643-160862643 on Build GRCh38
3477042 c.19A>T p.T7S Chr.1: 160862659-160862659 on Build GRCh38
4518778 c.7_8GG>AA p.G3K Chr.1: 160862670-160862671 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374829810 Chr.1: 160862645-160862645 on Build GRCh38
rs769228484 Chr.1: 160862662-160862662 on Build GRCh38
rs908450567 Chr.1: 160862627-160862627 on Build GRCh38
rs149914624 Chr.1: 160862653-160862653 on Build GRCh38
rs146428268 Chr.1: 160862673-160862673 on Build GRCh38
rs982621630 Chr.1: 160862621-160862621 on Build GRCh38
rs771344545 Chr.1: 160862638-160862638 on Build GRCh38
rs151127237 Chr.1: 160862666-160862666 on Build GRCh38
rs756637915 Chr.1: 160862705-160862705 on Build GRCh38
rs753343734 Chr.1: 160862670-160862670 on Build GRCh38
rs773214095 Chr.1: 160862609-160862609 on Build GRCh38
rs189395221 Chr.1: 160862694-160862694 on Build GRCh38
rs866413602 Chr.1: 160862619-160862619 on Build GRCh38
rs16832486 Chr.1: 160862738-160862738 on Build GRCh38
rs566233906 Chr.1: 160862657-160862657 on Build GRCh38
rs199618379 Chr.1: 160862597-160862597 on Build GRCh38
rs771074498 Chr.1: 160862713-160862713 on Build GRCh38
rs755368778 Chr.1: 160862690-160862690 on Build GRCh38
rs768190241 Chr.1: 160862611-160862612 on Build GRCh38
rs746044600 Chr.1: 160862727-160862727 on Build GRCh38
rs749382682 Chr.1: 160862708-160862708 on Build GRCh38
rs770305568 Chr.1: 160862603-160862603 on Build GRCh38
rs16832484 Chr.1: 160862639-160862639 on Build GRCh38
rs778536202 Chr.1: 160862595-160862595 on Build GRCh38
rs368378136 Chr.1: 160862737-160862737 on Build GRCh38
rs766128335 Chr.1: 160862732-160862732 on Build GRCh38
rs115596289 Chr.1: 160862699-160862699 on Build GRCh38
rs767166829 Chr.1: 160862656-160862656 on Build GRCh38
rs774734877 Chr.1: 160862716-160862716 on Build GRCh38
rs1055935541 Chr.1: 160862680-160862680 on Build GRCh38
rs372332184 Chr.1: 160862592-160862592 on Build GRCh38
rs369932468 Chr.1: 160862599-160862599 on Build GRCh38
rs749366273 Chr.1: 160862611-160862611 on Build GRCh38
rs764885317 Chr.1: 160862674-160862674 on Build GRCh38
rs374543049 Chr.1: 160862583-160862583 on Build GRCh38
rs753212768 Chr.1: 160862723-160862724 on Build GRCh38
rs369152438 Chr.1: 160862683-160862683 on Build GRCh38
rs770608483 Chr.1: 160862731-160862731 on Build GRCh38
rs747382923 Chr.1: 160862596-160862596 on Build GRCh38
rs760567350 Chr.1: 160862663-160862663 on Build GRCh38
rs911134142 Chr.1: 160862560-160862560 on Build GRCh38
rs186849603 Chr.1: 160862589-160862589 on Build GRCh38
rs778338734 Chr.1: 160862707-160862707 on Build GRCh38
rs753354373 Chr.1: 160862590-160862590 on Build GRCh38

View Full Product Details