Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48550782-48551015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXB-AS1
Entrez Gene ID
100874362
Gene Name
HOXB cluster antisense RNA 1
Gene Aliases
-
UniGene
Hs.171806
Gene Chromosome Location
Chr.17: 48544351-48551241 on Build GRCh38
Transcripts
NR_102279.1 NM_002146.4 XM_005257277.3 XM_005257280.3 XM_005257282.3 XM_006721854.2 XM_011524707.2 XM_011524708.2 XM_011524710.1 XM_011524719.1 XM_011524720.2 XM_011524721.2 XM_011524726.2 XM_017024557.1 XM_017024558.1 XM_017024559.1 XM_017024560.1 XM_017024561.1 XM_017024562.1 XM_017024563.1

Gene Symbol
HOXB3
Entrez Gene ID
3213
Gene Name
homeobox B3
Gene Aliases
HOX2, HOX2G, Hox-2.7
UniGene
Hs.654560
Gene Chromosome Location
Chr.17: 48548870-48590369 on Build GRCh38
Transcripts
NR_102279.1 NM_002146.4 XM_005257277.3 XM_005257280.3 XM_005257282.3 XM_006721854.2 XM_011524707.2 XM_011524708.2 XM_011524710.1 XM_011524719.1 XM_011524720.2 XM_011524721.2 XM_011524726.2 XM_017024557.1 XM_017024558.1 XM_017024559.1 XM_017024560.1 XM_017024561.1 XM_017024562.1 XM_017024563.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1254353 c.764C>A p.S255* Chr.17: 48550866-48550866 on Build GRCh38
6681684 c.685G>A p.E229K Chr.17: 48550945-48550945 on Build GRCh38
4887871 c.650G>A p.R217H Chr.17: 48550980-48550980 on Build GRCh38
4802664 c.761C>T p.S254L Chr.17: 48550869-48550869 on Build GRCh38
5852826 c.643C>T p.R215W Chr.17: 48550987-48550987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756366667 Chr.17: 48550879-48550879 on Build GRCh38
rs200719688 Chr.17: 48550815-48550815 on Build GRCh38
rs201674142 Chr.17: 48550835-48550835 on Build GRCh38
rs569979335 Chr.17: 48550934-48550934 on Build GRCh38
rs762955717 Chr.17: 48550832-48550833 on Build GRCh38
rs910527226 Chr.17: 48550854-48550854 on Build GRCh38
rs780233987 Chr.17: 48550824-48550824 on Build GRCh38
rs200298834 Chr.17: 48550844-48550844 on Build GRCh38
rs770510894 Chr.17: 48550937-48550937 on Build GRCh38
rs373574756 Chr.17: 48550885-48550885 on Build GRCh38
rs563790203 Chr.17: 48550881-48550881 on Build GRCh38
rs753275502 Chr.17: 48550852-48550852 on Build GRCh38
rs368031900 Chr.17: 48550866-48550866 on Build GRCh38
rs751396549 Chr.17: 48550895-48550897 on Build GRCh38
rs199720030 Chr.17: 48550862-48550862 on Build GRCh38
rs766032924 Chr.17: 48550856-48550857 on Build GRCh38
rs750633092 Chr.17: 48550871-48550871 on Build GRCh38
rs778112969 Chr.17: 48550810-48550810 on Build GRCh38
rs142877220 Chr.17: 48550857-48550857 on Build GRCh38
rs867448391 Chr.17: 48550915-48550915 on Build GRCh38
rs146112353 Chr.17: 48550861-48550861 on Build GRCh38
rs35517955 Chr.17: 48550817-48550817 on Build GRCh38
rs761508855 Chr.17: 48550834-48550834 on Build GRCh38
rs746004710 Chr.17: 48550816-48550816 on Build GRCh38
rs201126398 Chr.17: 48550814-48550814 on Build GRCh38
rs538887217 Chr.17: 48550949-48550949 on Build GRCh38
rs762333689 Chr.17: 48550986-48550986 on Build GRCh38
rs749271465 Chr.17: 48550828-48550828 on Build GRCh38
rs772713771 Chr.17: 48550838-48550838 on Build GRCh38
rs759045256 Chr.17: 48550958-48550958 on Build GRCh38
rs771782606 Chr.17: 48550922-48550922 on Build GRCh38
rs768655705 Chr.17: 48550829-48550829 on Build GRCh38
rs764629687 Chr.17: 48550963-48550963 on Build GRCh38
rs781526391 Chr.17: 48550868-48550868 on Build GRCh38
rs774344428 Chr.17: 48550833-48550833 on Build GRCh38
rs758859201 Chr.17: 48550809-48550809 on Build GRCh38
rs748024750 Chr.17: 48550914-48550914 on Build GRCh38
rs148521405 Chr.17: 48550845-48550845 on Build GRCh38
rs923515927 Chr.17: 48550886-48550886 on Build GRCh38
rs369506276 Chr.17: 48550825-48550825 on Build GRCh38
rs774703442 Chr.17: 48550964-48550964 on Build GRCh38
rs768778823 Chr.17: 48550897-48550897 on Build GRCh38
rs749364149 Chr.17: 48550889-48550889 on Build GRCh38
rs149347321 Chr.17: 48550811-48550811 on Build GRCh38
rs778683908 Chr.17: 48550899-48550899 on Build GRCh38
rs760322891 Chr.17: 48550843-48550843 on Build GRCh38
rs767804647 Chr.17: 48550987-48550987 on Build GRCh38

View Full Product Details