Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 86265942-86266183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EED
Entrez Gene ID
8726
Gene Name
embryonic ectoderm development
Gene Aliases
HEED, WAIT1
UniGene
Hs.503510
Gene Chromosome Location
Chr.11: 86244384-86285420 on Build GRCh38
Transcripts
NM_001308007.1 NM_003797.4 XM_005274373.2 XM_011545330.1 XM_011545331.1 XM_017018512.1 XM_017018513.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
691224 c.788G>T p.W263L Chr.11: 86266144-86266144 on Build GRCh38
4507840 c.758C>T p.S253F Chr.11: 86266114-86266114 on Build GRCh38
3792004 c.778C>T p.L260F Chr.11: 86266134-86266134 on Build GRCh38
6365110 c.793A>G p.I265V Chr.11: 86266149-86266149 on Build GRCh38
6935405 c.791G>T p.R264M Chr.11: 86266147-86266147 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs972622503 Chr.11: 86266072-86266072 on Build GRCh38
rs766054344 Chr.11: 86266052-86266052 on Build GRCh38
rs772462856 Chr.11: 86266065-86266065 on Build GRCh38
rs777367733 Chr.11: 86265971-86265971 on Build GRCh38
rs577614545 Chr.11: 86266127-86266127 on Build GRCh38
rs769990056 Chr.11: 86266115-86266115 on Build GRCh38
rs938322484 Chr.11: 86266070-86266070 on Build GRCh38
rs74429367 Chr.11: 86265990-86265990 on Build GRCh38
rs1017647245 Chr.11: 86265970-86265970 on Build GRCh38
rs759894228 Chr.11: 86266094-86266094 on Build GRCh38
rs909385286 Chr.11: 86266023-86266023 on Build GRCh38
rs912218179 Chr.11: 86266011-86266011 on Build GRCh38
rs746480957 Chr.11: 86265976-86265976 on Build GRCh38
rs748653059 Chr.11: 86266057-86266057 on Build GRCh38
rs950922737 Chr.11: 86266004-86266004 on Build GRCh38
rs926983041 Chr.11: 86266067-86266067 on Build GRCh38
rs371131577 Chr.11: 86266077-86266077 on Build GRCh38
rs751212980 Chr.11: 86266064-86266064 on Build GRCh38
rs10898460 Chr.11: 86266075-86266075 on Build GRCh38
rs983545744 Chr.11: 86266018-86266018 on Build GRCh38
rs538972736 Chr.11: 86265991-86265991 on Build GRCh38
rs755601508 Chr.11: 86266035-86266035 on Build GRCh38
rs976212559 Chr.11: 86265972-86265972 on Build GRCh38
rs779601207 Chr.11: 86266036-86266036 on Build GRCh38
rs775716311 Chr.11: 86266147-86266147 on Build GRCh38

View Full Product Details