Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46648688-46648929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DACT3
Entrez Gene ID
147906
Gene Name
dishevelled binding antagonist of beta catenin 3
Gene Aliases
DAPPER3, RRR1
UniGene
Hs.515490
Gene Chromosome Location
Chr.19: 46647564-46661138 on Build GRCh38
Transcripts
NM_001301046.1 NM_145056.2 XM_011526499.2 XM_011526500.2 XM_017026334.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1590177 c.1649C>T p.S550L Chr.19: 46648723-46648723 on Build GRCh38
4876375 c.1660G>A p.E554K Chr.19: 46648712-46648712 on Build GRCh38
1659328 c.1572C>A p.C524* Chr.19: 46648800-46648800 on Build GRCh38
5730383 c.923G>T p.G308V Chr.19: 46648774-46648774 on Build GRCh38
6368582 c.1613C>T p.A538V Chr.19: 46648759-46648759 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs958081299 Chr.19: 46648783-46648783 on Build GRCh38
rs1055848954 Chr.19: 46648797-46648797 on Build GRCh38
rs752932724 Chr.19: 46648743-46648743 on Build GRCh38
rs552073626 Chr.19: 46648728-46648728 on Build GRCh38
rs58806479 Chr.19: 46648830-46648830 on Build GRCh38
rs1035991464 Chr.19: 46648856-46648856 on Build GRCh38
rs552110358 Chr.19: 46648772-46648772 on Build GRCh38
rs930353192 Chr.19: 46648872-46648872 on Build GRCh38
rs774790938 Chr.19: 46648740-46648740 on Build GRCh38
rs894526552 Chr.19: 46648837-46648837 on Build GRCh38
rs747697989 Chr.19: 46648719-46648719 on Build GRCh38
rs776228869 Chr.19: 46648714-46648714 on Build GRCh38
rs908471324 Chr.19: 46648718-46648718 on Build GRCh38
rs767881568 Chr.19: 46648742-46648742 on Build GRCh38
rs898926309 Chr.19: 46648868-46648868 on Build GRCh38
rs1009260109 Chr.19: 46648905-46648905 on Build GRCh38
rs1031046597 Chr.19: 46648773-46648774 on Build GRCh38
rs766728501 Chr.19: 46648738-46648738 on Build GRCh38
rs764216094 Chr.19: 46648749-46648749 on Build GRCh38
rs559821581 Chr.19: 46648784-46648784 on Build GRCh38
rs768322728 Chr.19: 46648712-46648712 on Build GRCh38
rs760016904 Chr.19: 46648900-46648900 on Build GRCh38
rs780873448 Chr.19: 46648780-46648780 on Build GRCh38
rs754738498 Chr.19: 46648775-46648775 on Build GRCh38
rs898355155 Chr.19: 46648763-46648763 on Build GRCh38
rs753797793 Chr.19: 46648762-46648762 on Build GRCh38
rs1050705171 Chr.19: 46648883-46648883 on Build GRCh38
rs940338091 Chr.19: 46648854-46648854 on Build GRCh38
rs927652781 Chr.19: 46648851-46648851 on Build GRCh38
rs774957424 Chr.19: 46648885-46648885 on Build GRCh38
rs978181704 Chr.19: 46648754-46648754 on Build GRCh38
rs769256147 Chr.19: 46648788-46648788 on Build GRCh38
rs769273593 Chr.19: 46648723-46648723 on Build GRCh38
rs779809717 Chr.19: 46648765-46648765 on Build GRCh38
rs559668585 Chr.19: 46648861-46648861 on Build GRCh38
rs759929125 Chr.19: 46648741-46648741 on Build GRCh38
rs948326198 Chr.19: 46648826-46648826 on Build GRCh38
rs780113189 Chr.19: 46648768-46648768 on Build GRCh38
rs758372814 Chr.19: 46648764-46648764 on Build GRCh38
rs756307687 Chr.19: 46648748-46648748 on Build GRCh38
rs772782434 Chr.19: 46648798-46648798 on Build GRCh38
rs771489153 Chr.19: 46648871-46648871 on Build GRCh38
rs184698514 Chr.19: 46648737-46648737 on Build GRCh38
rs982371161 Chr.19: 46648841-46648841 on Build GRCh38

View Full Product Details