Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100390643-100390914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC14A
Entrez Gene ID
8556
Gene Name
cell division cycle 14A
Gene Aliases
DFNB105, cdc14, hCDC14
UniGene
Hs.127411
Gene Chromosome Location
Chr.1: 100345015-100520281 on Build GRCh38
Transcripts
NM_001319210.1 NM_001319211.1 NM_001319212.1 NM_003672.3 NM_033312.2 NM_033313.2 XM_005271294.3 XM_005271296.3 XM_011542340.2 XM_011542341.2 XM_011542345.2 XM_011542346.2 XM_017002646.1 XM_017002647.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4670597 c.277G>A p.A93T Chr.1: 100390792-100390792 on Build GRCh38
892969 c.233G>T p.R78I Chr.1: 100390748-100390748 on Build GRCh38
1738517 c.268C>T p.R90W Chr.1: 100390783-100390783 on Build GRCh38
892970 c.249C>A p.H83Q Chr.1: 100390764-100390764 on Build GRCh38
5090467 c.237A>G p.R79R Chr.1: 100390749-100390749 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754485754 Chr.1: 100390877-100390877 on Build GRCh38
rs757163599 Chr.1: 100390673-100390673 on Build GRCh38
rs753571855 Chr.1: 100390853-100390853 on Build GRCh38
rs770059150 Chr.1: 100390790-100390790 on Build GRCh38
rs372163710 Chr.1: 100390696-100390696 on Build GRCh38
rs762955081 Chr.1: 100390804-100390804 on Build GRCh38
rs758333044 Chr.1: 100390885-100390885 on Build GRCh38
rs28361212 Chr.1: 100390824-100390824 on Build GRCh38
rs777222568 Chr.1: 100390712-100390713 on Build GRCh38
rs200908835 Chr.1: 100390692-100390692 on Build GRCh38
rs374536334 Chr.1: 100390724-100390724 on Build GRCh38
rs971761358 Chr.1: 100390675-100390675 on Build GRCh38
rs762571830 Chr.1: 100390717-100390718 on Build GRCh38
rs755875156 Chr.1: 100390747-100390747 on Build GRCh38
rs919023543 Chr.1: 100390677-100390677 on Build GRCh38
rs771875274 Chr.1: 100390782-100390782 on Build GRCh38
rs1054058414 Chr.1: 100390773-100390773 on Build GRCh38
rs779708565 Chr.1: 100390750-100390750 on Build GRCh38
rs779794065 Chr.1: 100390827-100390827 on Build GRCh38
rs1035897020 Chr.1: 100390822-100390822 on Build GRCh38
rs143209870 Chr.1: 100390766-100390766 on Build GRCh38
rs748084346 Chr.1: 100390883-100390883 on Build GRCh38
rs1002702385 Chr.1: 100390821-100390821 on Build GRCh38
rs375521987 Chr.1: 100390712-100390712 on Build GRCh38
rs906650468 Chr.1: 100390820-100390820 on Build GRCh38
rs750068985 Chr.1: 100390734-100390734 on Build GRCh38
rs374836642 Chr.1: 100390838-100390838 on Build GRCh38
rs759797751 Chr.1: 100390784-100390784 on Build GRCh38
rs544984501 Chr.1: 100390719-100390719 on Build GRCh38
rs772919579 Chr.1: 100390783-100390783 on Build GRCh38
rs199907180 Chr.1: 100390791-100390791 on Build GRCh38
rs778332362 Chr.1: 100390879-100390879 on Build GRCh38
rs140021980 Chr.1: 100390817-100390817 on Build GRCh38
rs369158285 Chr.1: 100390761-100390761 on Build GRCh38
rs146977673 Chr.1: 100390819-100390819 on Build GRCh38
rs764190305 Chr.1: 100390810-100390810 on Build GRCh38
rs748056301 Chr.1: 100390778-100390778 on Build GRCh38
rs376488789 Chr.1: 100390758-100390758 on Build GRCh38
rs368310370 Chr.1: 100390721-100390721 on Build GRCh38
rs145199717 Chr.1: 100390755-100390755 on Build GRCh38
rs757602119 Chr.1: 100390718-100390718 on Build GRCh38
rs752058562 Chr.1: 100390812-100390812 on Build GRCh38

View Full Product Details